Muffle Furnaces
Thermo Scientific™ Lindberg/Blue M™ Moldatherm™ Box Furnaces Available on GSA/VA Contract for Federal Government customers only.
Minimize outer surface temperatures while maintaining uniform heat distribution within the chamber. Suitable for a variety of industrial and laboratory applications.
Thermo Scientific™ Thermolyne™ Industrial Benchtop Muffle Furnaces Available on GSA/VA Contract for Federal Government customers only.
Perform ultra-high temperature applications with this industrial benchtop muffle furnace, which reaches a maximum temperature of 1200°C.
Thermo Scientific™ Thermolyne™ Largest Tabletop Muffle Furnaces Available on GSA/VA Contract for Federal Government customers only.
Triple the usable working area with two supplied accessory refractory shelves using Thermolyne Largest Tabletop Muffle Furnace. Available in three temperature control models.
Thermo Scientific™ Thermolyne™ Benchtop 1100°C Muffle Furnaces Available on GSA/VA Contract for Federal Government customers only.
Reduce energy consumption and increase heatup time with Thermo Scientific™ Thermolyne™ Benchtop 1100°C  Muffle Furnaces.
Thermo Scientific™ Thermolyne™ Premium Large Muffle Furnaces Available on GSA/VA Contract for Federal Government customers only.
Conduct industrial applications including ashing organic and inorganic samples and gravimetric analysis with Thermo Scientific™ Thermolyne™ Premium Large Muffle Furnaces.
Thermo Scientific™ Lindberg/Blue M™ LGO Box Furnaces Available on GSA/VA Contract for Federal Government customers only.
Utilize the latest technical advances in heating elements, insulation and temperature control integrated into a self-contained cabinet with these box furnaces.
Thermo Scientific™ Lindberg/Blue M™ 1700°C Box Furnaces, Integral Control Available on GSA/VA Contract for Federal Government customers only.
Heat to high temperatures faster using Lindberg/Blue M 1700°C furnaces. Designed for efficient, high-temperature use with minimal maintenance.
Thermo Scientific™ Lindberg/Blue M™ 1700°C Box Furnaces Available on GSA/VA Contract for Federal Government customers only.
Heat to high temperatures faster with Thermo Scientific Lindberg/Blue M 1700 C furnaces. They are designed for efficient, high-temperature use with minimal maintenance.
Thermo Scientific™ Lindberg/Blue M™ Multipurpose 1500°C Box Furnaces Available on GSA/VA Contract for Federal Government customers only.
Lindberg/Blue M multipurpose furnaces feature integral control to 1500°C. Double-wall insulated construction provides rapid heatup/cooldown and energy efficiency.
Thermo Scientific™ Lindberg/Blue M™ Heavy-Duty 1200°C Box Furnaces Available on GSA/VA Contract for Federal Government customers only.
Rapidly heat, recover and cool with the outstanding temperature uniformity of Thermo Scientific™ Lindberg/Blue M™ Heavy-Duty 1200°C Box Furnaces.
Thermo Scientific™ Muffle Furnaces Products for Science Education
Ideal for the determination of volatile and suspended solids, drying and evaporating, ignition tests, gravimetric analysis, ashing organic and inorganic samples and heat treatment operations.
  spinner