Heater Accessories
Jelliff Wire Gauze
Chrome and iron wire gauze
Eisco Ceramic-Centered Iron Wire Gauze Products for Science Education
Eisco Iron Wire Gauze with Ceramic Center is high quality and extra strong for long-term use in the classroom laboratory.
United Scientific Supplies Wire Gauzes with Ceramic Center Products for Science Education
Used for general heating purposes in laboratories.
GSC Go Science Crazy Wire Gauze Products for Science Education
Aids in the distribution of heat while using a Bunsen burner. GSC International Wire Gauze features rolled smooth wire edges. A variety of sizes are available.
EISCO Wire Gauge Products for Science Education
For measuring the diameter of wire.
GSC Go Science Crazy Wire Gauze with Ceramic Center Products for Science Education
Distributes heat evenly while using a Bunsen burner. GSC International Wire Gauze with Ceramic Center features rolled smooth wire edges and a clean white center ceramic. A variety of sizes are available.
United Scientific Supplies Wire Gauzes, Plain Products for Science Education
Used for general heating purposes in laboratories.
American Educational Products Durable Gauze Products for Science Education
With ceramic fiber centers for thermal protection.
American Educational Products Iron Wire Gauze Products for Science Education
Economical, general-purpose wire gauze — a laboratory staple.
Nickel-Chromium Wire Gauze Products for Science Education
Withstands a hotter flame than iron gauze.
  spinner