Process Moisture Analyzers
Sartorius™ Heater Module (1mg, 40g) for Mark 3 LTE Moisture Analyzer
Modular add-on to the Sartorius Mark 3 LTE Moisture Analyzer (max. two heater modules). System status indicator lights include two tri-color LEDs (balance and heater) on the front of the heater module.
Sartorius™ LMA200PM Microwave Moisture Analyzer
Produces results up to 300x faster than a drying oven for samples with high moisture content. Sartorius LMA200PM Microwave Moisture Analyzer has an in-built analytical balance which ensures high precision in moisture analysis.
  spinner