Hollow Cathode Lamps and Light Sources
Fisherbrand™ Hollow Cathode Lamps
Window materials are quartz, borosilicate and UV glass; Lamps are single-element and 1 1/2 in. (37mm) in diameter
Fisherbrand™ Hollow Cathode Multi-Element 1 1/2in. Lamps
Window materials are quartz, borosilicate and UV glass; Lamps are multi-element and 1 1/2 in. (37mm) in diameter
Fisherbrand™ Hollow Cathode Single-Element 2in. dia. Lamps
Window materials are quartz, borosilicate and UV glass; Lamps are single-element and 2 in. diameter
Fisherbrand™ Hollow Cathode Multi-Element 2in. Lamps
Window materials are quartz, borosilicate and UV glass; Lamps are multi-element and 2 in. diameter
  spinner