Photoionization Lamps
Restek™ Photoionization Lamps
Higher output makes it ideal for detection of BTEX compounds.
Fisherbrand™ Photoionization Lamps
Available in the DC style in the 10.0 to 10.6eV range commonly used for most analyses
Restek™ Polishing Kit for Photoionization Lamps
Used to clean the outside of the PID lamp window to restore the output.
  spinner