Ultraviolet Lamps and Accessories
Thermo Scientific™ Pierce™ UVP 3UV Ultraviolet Lamp
Photoactivate crosslinkers in UV-crosslinking methods with these compact, UV-lamps with 252nm, 302nm and 365nm short to long wavelength bulbs.
UVP Compact and Handheld UV Lamps
Essential multipurpose UV lamps for laboratory use
Spectroline™ UV Darkroom Cabinet
Portable, space-saving mini-cabinet doubles as a personal darkroom and UV irradiating units
EMD Millipore™ 254nm UV Lamp for Thin-Layer Chromatography Available on GSA/VA Contract for Federal Government customers only.
In contrast to lamps with wavelength filters and very narrow spectrum, this lamp has a broader spectrum for quick analyses or screening purposes
Spectroline™ Germicidal UV Sterilizing Lamps
Unfiltered 254nm shortwave (UV-C) lamps provide high-intensity, wide area UV sterilization coverage
UVP Blak-Ray™ B-100A UV Lamps
High powered lamps offer brilliant irradiance for optimum fluorescence
UVP Compact and Handheld UV Lamp Replacement Tubes
For use with 4w and 6w UV lamps.
UVP UVLS-26 EL Series UV 6Watt Lamp 115V
Light weight, yet durable for high usage
UVP XX-Series UV Bench Lamp, 115V
Bench lamps that offer a choice between longwave, shortwave or midrange wavelengths for use in sterilization/sanitation and fluorescence/inspection applications. UVP XX-Series UV Bench Lamp, 115V are designed for use in the United States.
UVP UVL-26 EL Series 6-Watt UV Lamp
Light weight, yet durable for high usage
Spectroline™ MiniMAX™ UV Lamps
Compact, lightweight and portable for site or field inspections
Spectroline™ E-Series Handheld Lamps
A full range of models in this lamp series
UVP Blak-Ray™ B-100AP High-Intensity UV Inspection Lamps: UV Lamp with 20 ft. Cord
Accessory for B-100AP High-Intensity UV Inspection Lamps
UVP High-Performance Mineralight™ Display Lamps: Replacement Tubes
UVP 25-watt tubes providing hours of UV illumination when used with High-Performance Mineralight Display Lamps.
UVP EL Series 8W UV Lamps
Durable, scratch-resistant, lightweight and versatile
UVP Blak-Ray™ UV Benchtop Lamps
Longwave (365nm) bench lamps are available in selection of 15, 20 and 40-watt housings
UVP Mini UV Lamps
Battery operated and used as portable lamp for industrial, laboratory, lapidary and field inspections.
UVP UVP XX-15 Exposure Stand
For use with UVP XX-15 UV Lamps
UVP Pen-Ray™ Field Pack
Includes a Pen-Ray™ UV light source and a DC power supply (with 9V battery) in a compact, portable package
UVP XX-Series UV Bench Lamp Replacement Tubes
UVP™ XX-Series UV Bench Lamp Replacement Tubes are interchangeable UV tubes available in shortwave, longwave and mid-range wavelengths.
UVP Replacement UV Light Tubes
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
UVP Mini Ultraviolet Lamp Replacement Tube
Replacement tubes for lightweight, compact and financial UV Lamps
UVP UVL-18 EL Series 8-Watt Lamp
Light weight, rugged design is excellent for high usage
UVP Standard Mercury Pen-Ray Models
Low-pressure mercury gaseous discharge handheld lamps are ideal for small apertures requiring spectroscopic wavelength calibration, sterilization, UV curing or fluorescence detection applications
  spinner