Pipetting Workstations
Eppendorf™ Repeater™ M4 Manual Handheld Pipette Dispenser Available on GSA/VA Contract for Federal Government customers only.
Uses positive displacement principal to precisely and accurately dispense virtually any liquid. Eppendorf™ Repeater™ M4 Manual Handheld Pipette Dispenser dispenses liquid directly without air cushion, ensuring correct volume is dispensed regardless of density, viscosity and volatility of liquid.
Thermo Scientific™ Versette™ Pipetting Heads
Choose from a volume range of 0.5μL to 300μL with Thermo Scientific™ Versette™ Pipetting Heads.
BioTek™ MultiFlo™ Microplate Dispenser Available on GSA/VA Contract for Federal Government customers only.
Replaces up to four independent dispensers in one compact instrument
Neutec Group BioTool Swiss Dosi-Pump DP1000 Agar and Liquid Pump
State-of-the-art technology is suitable for various applications for liquid dispensing and agar filling
Neutec Group Eddy Jet™ 2 Spiral Plating System
For spiral inoculation with disposable syringes
  spinner