Antibody Production and Purification Reagents and Kits
BD Cytometric Bead Array Phospho p38 (T180/Y182) Standard Available on GSA/VA Contract for Federal Government customers only.
Lyophilized phosphorylated recombinant p38
BD Cytometric Bead Array Human Interleukin-13 Flex Set Standard Available on GSA/VA Contract for Federal Government customers only.
Reconstituted Human IL-13 Standard is useful as a standard control with the CBA Human IL-13 Flex Set
BD Cytometric Bead Array Rat TNF Flex Set Standard Available on GSA/VA Contract for Federal Government customers only.
Useful as a standard control with CBA Rat TNF Flex Set
BD Cytometric Bead Array Human MIP-1α Standard Available on GSA/VA Contract for Federal Government customers only.
Useful as a standard control with the CBA Human MIP-1a Flex Set
BD Cytometric Bead Array Phospho JNK1/2 (T183/Y185) Standard Available on GSA/VA Contract for Federal Government customers only.
Lyophilized recombinant Phospho JNK1/2 (T183/Y185)
BD Cytometric Bead Array Mouse IL-4 Flex Set Standard Available on GSA/VA Contract for Federal Government customers only.
Standard control for flow cytometry
BD Cytometric Bead Array Mouse Th1/Th2 Cytokine Standard Available on GSA/VA Contract for Federal Government customers only.
Designed for use as a standard control with the Cytometric Bead Array (CBA) Mouse Th1/Th2 Cytokine Kit
BD Cytometric Bead Array Phospho ERK1/2 (T202/Y204) Standard Available on GSA/VA Contract for Federal Government customers only.
Lyophilized phosphorylated recombinant extracellular signal regulated kinases 1/2
BD Cytometric Bead Array Human MCP-1 Standard Available on GSA/VA Contract for Federal Government customers only.
Useful as a standard control with the CBA Human MCP-1 Flex Set
BD Cytometric Bead Array Mouse IL-2 Flex Set Standard Available on GSA/VA Contract for Federal Government customers only.
Standard control for flow cytometry
BD Cytometric Bead Array Mouse IFN-γ Flex Set Standard Available on GSA/VA Contract for Federal Government customers only.
Useful as a standard control with CBA Mouse IFN-γ Flex Set
GE Healthcare HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
Designed for method optimization and parameter screening for capture of monoclonal antibodies.   GE Healthcare HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns come ready-to-use  for convenience.
Gibson Bioscience Reagents: Kovacs
For the detection of microorganisms that produce Indole as a by-product of their metabolism
Gibson Bioscience Reagents: Ehrlichs
Use as an indicator reagent for the indole test for gram-negative bacteria producing minute quantities of indole
Gibson Bioscience Reagents: Nitrate A
For detection of the reduction of nitrate to nitrite
Gibson Bioscience Reagents: PYR
For use with PYR and LAP disks
Gibson Bioscience Reagents: Ferric Chloride 10%
Used as a color developer for phenylalanine agar tests and tryptophan deaminase tests
Gibson Bioscience Reagents: KOH 3%
Solubility test to aid in gram stain reactions
Gibson Bioscience Reagents: Catalese
For the detection of microorganisms that possess the catalase enzyme
  spinner