Primary Antibodies Hg to Hi
HIST2H2AC Polyclonal Antibody, Invitrogen™
HIST2H2AC Antibody, PA5-43691, from Invitrogen™. Species Reactivity: Bovine, Canine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat; Applications: Western Blot.
HIP1 Polyclonal Antibody, Invitrogen™
HIP1 Antibody, PA5-46708, from Invitrogen™. Species Reactivity: Human; Applications: Western Blot.
Phospho-Histone H3 pSer28 Polyclonal Antibody, Invitrogen™
Phospho-Histone H3 pSer28 Antibody, PA5-17318, from Invitrogen™. Species Reactivity: Bv, Ck, Ds, Hm, Hu, Ms, Rt, Xnl, Zb; Applications: FACS, ICC, IF, IP, WB.
Hic5 Mouse, Unlabeled, Clone: 34, BD Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
HIV1 gp41 HRP Polyclonal Antibody, Invitrogen™
HIV1 gp41 Antibody, HRP conjugate, PA1-73099, from Invitrogen™. Species Reactivity: Virus; Applications: ELISA, Western Blot.
HIST2H2AA3 Polyclonal Antibody, Invitrogen™
HIST2H2AA3 Antibody, PA5-43692, from Invitrogen™. Species Reactivity: Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat; Applications: Western Blot.
HIST1H2BM Polyclonal Antibody, Invitrogen™
HIST1H2BM Antibody, PA5-49358, from Invitrogen™. Species Reactivity: Human; Applications: Western Blot.
Histamine Polyclonal Antibody, Invitrogen™
Histamine Antibody, PA1-36089, from Invitrogen™. Species Reactivity: Rat; Applications: Immunohistochemistry (Paraffin, Frozen).
HIPK4 Polyclonal Antibody, Invitrogen™
HIPK4 Antibody, PA5-43886, from Invitrogen™. Species Reactivity: Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Rabbit, Rat; Applications: Western Blot.
HGK/MAP4K4 Polyclonal Antibody, Invitrogen™
HGK/MAP4K4 Antibody, PA5-49780, from Invitrogen™. Species Reactivity: Human, Mouse; Applications: ICC, IF, WB.
Phospho-HIPK2 pTyr361 Polyclonal Antibody, Invitrogen™
Phospho-HIPK2 pTyr361 Antibody, PA5-13045, from Invitrogen™. Species Reactivity: Human; Applications: Dot blot.
HIV1 p24 Monoclonal Antibody, Clone: 5 (05-001), Invitrogen™
HIV1 p24 Antibody (5 (05-001)), MA1-71515, from Invitrogen™. Species Reactivity: Virus; Applications: ELISA, Immunofluorescence, Western Blot.
6x-His Epitope Tag Monoclonal Antibody, Clone: 13/45/31-2, Invitrogen™
6x-His Epitope Tag Antibody (13/45/31-2), MA1-4806, from Invitrogen™. Species Reactivity: Tag; Applications: ELISA, FACS, IF, IHC (Frozen), IP, WB.
HIV1 gp41 Biotin Polyclonal Antibody, Invitrogen™
HIV1 gp41 Antibody, Biotin conjugate, PA1-73100, from Invitrogen™. Species Reactivity: Virus; Applications: Western Blot.
HIV1 Tat Monoclonal Antibody, Clone: 15 (Tat 15), Invitrogen™
HIV1 Tat Antibody (15 (Tat 15)), MA1-71510, from Invitrogen™. Species Reactivity: Virus; Applications: ChIP assay, ELISA, Western Blot.
Histidine Decarboxylase Polyclonal Antibody, Invitrogen™
Histidine Decarboxylase Antibody, PA1-36090, from Invitrogen™. Species Reactivity: Canine, Guinea Pig, Human, Mouse, Rat; Applications: IF, IHC (Paraffin, Frozen).
6x-His Epitope Tag Monoclonal Antibody, Clone: HIS.H8, Invitrogen™
6x-His Epitope Tag Antibody (HIS.H8), MA1-21315, from Invitrogen™. Species Reactivity: Tag; Applications: ELISA, ICC, IF, IP, WB.
HINT1 Polyclonal Antibody, Invitrogen™
HINT1 Antibody, PA5-20613, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunocytochemistry, Western Blot.
G-protein coupled receptor homolog U12 Polyclonal Antibody, Invitrogen™
G-protein coupled receptor homolog U12 Antibody, PA5-39863, from Invitrogen™. Species Reactivity: Virus; Applications: Western Blot.
  spinner