Biochemicals and Reagents
Thermo Scientific™ dNTP Mixes 
Reduce the number of pipetting steps and the risk of reaction setup errors with these dNTP mixes, premixed aqueous solutions of dATP, dTTP, dCTP, and dGTP.
Thermo Scientific™ Phusion High-Fidelity DNA Polymerase 
Get superior performance for cloning and other applications requiring high fidelity with one of the most accurate thermostable DNA polymerases available.
Thermo Scientific™ GeneJET™ Plasmid Miniprep Kit 
Perform isolation of high quality plasmid DNA from recombinant E. coli cultures with this simple, rapid, and cost-effective system.
Thermo Scientific™ GeneRuler™ Ready-to-use 1kb Plus DNA Ladder 
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.
Thermo Scientific™ Phusion™ Hot Start II High-Fidelity DNA Polymerase 
Optimize PCR with one of the most accurate hot start DNA polymerases on the market, featuring extreme specificity and improved performance.
Thermo Scientific™ GeneRuler™ 1kb Plus DNA Ladder 
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.
Thermo Scientific™ dNTP Set Promotion 
Optimize molecular biology applications with this dNTP set, consisting of aqueous solutions of dATP, dTTP, dCTP and dGTP supplied in separate tubes.
Thermo Scientific™ dNTP Set 
Optimize molecular biology applications with this dNTP set, consisting of aqueous solutions of dATP, dTTP, dCTP and dGTP supplied in separate tubes.
Thermo Scientific™ Phusion™ Flash High-Fidelity PCR Master Mix 
Perform short PCR protocols without compromising fidelity or yield with this master mix.
Thermo Scientific™ dATP 
Optimize molecular biology applications with this extremely pure dATP.
Thermo Scientific™ GeneRuler 100 bp DNA Ladder, Ready-to-Use 100 to 1000 bp 
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.
Thermo Scientific™ dNTP Mixes Promotion
Reduce the number of pipetting steps and the risk of reaction setup errors with these dNTP mixes, premixed aqueous solutions of dATP, dTTP, dCTP, and dGTP.
Invitrogen™ dNTP Mix (10 mM)
Mix of four nucleotides
Invitrogen™ TRIzol™ Reagent
Isolate RNA, DNA, and protein from the same sample
Applied Biosystems™ TaqMan™ Gene Expression Master Mix 
Precise and reliable real-time qPCR quantification over 9 logs of linear dynamic range with Applied Biosystems™ TaqMan™ Gene Expression Master Mix.
Invitrogen™ TOPO™ XL-2 Complete PCR Cloning Kit, with One Shot™ OmniMAX™2 T1R Chemically Competent E. coli 
Provides all the necessary elements for efficient cloning of extra-long PCR products (up to 13 kb)
Invitrogen™ Qubit™ dsDNA HS Assay Kit
Highly selective for double-stranded DNA (dsDNA) over RNA
Thermo Scientific™ PageRuler™ Plus Prestained 10-250kDa Protein Ladder 
Mixture of 9 blue-, orange- and green-stained proteins (10 to 250kDa) used as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Invitrogen™ RNaseZap™ RNase Decontamination Solution
Surface decontaminant that destroys RNases on contact
Invitrogen™ mMESSAGE mMACHINE™ T7 Transcription Kit
in vitro synthesis of large amounts of capped RNA
Invitrogen™ Lipofectamine™ 2000 Transfection Reagent
Versatile transfection reagent that has been shown to effectively transfect the widest variety of adherent and suspension cell lines
Thermo Scientific™ EZ-Link™ NHS-Desthiobiotin
Label intracellular proteins for affinity purification with streptavidin beads
Thermo Scientific™ Pierce™ Tetramethylethylenediamine (TEMED)
Efficiently catalyze polyacrylamide gel polymerization in SDS-PAGE using TEMED.
Invitrogen™ SYBR™ Safe™ DNA Gel Stain
Highly sensitive stain for visualization of DNA in agarose or acrylamide gels
Invitrogen™ Novex™ NuPAGE™ 4-12% Bis-Tris Protein Gels, 1.0mm, 12 well
Pre-cast polyacrylamide gels designed to give optimal separation of small- to medium-sized proteins under denaturing conditions
Thermo Scientific™ Verso™ cDNA synthesis Kit 
Perform robust transcription of RNA to create a complete cDNA pool.
Thermo Scientific™ SureTect™ Salmonella Species PCR Assay
Detect Salmonella species in even the most challenging food types with this multiplex real-time assay that provides highly-specific detection of the invA gene.