DNA Synthesis Reagents and Columns
Applied Biosystems™ Triethylamine Acetate (TEAA) 2.0 M Non-distribution product as customer accommodation.
This 2.0 M Triethylamine Acetate solution is used with OPC™ cartridges (Cat. No. 400771), for oligonucleotide purification.
Applied Biosystems™ Trap-Pak™ Bags (Mini) Non-distribution product as customer accommodation.
Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to reduce and maintain a low water content in solutions that must remain anhydrous.
Applied Biosystems™ Trap-Pak™ Bags (Medium) Non-distribution product as customer accommodation.
Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to reduce and maintain a low water content in solutions that must remain anhydrous.
Invitrogen™ GeneArt™ Seamless Cloning and Assembly Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Enables the simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute room temperature reaction
Invitrogen™ GeneArt™ Seamless Cloning and Assembly Enzyme Mix Non-distribution product as customer accommodation.
Enables the simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute room temperature reaction
Invitrogen™ GeneArt™ Seamless PLUS Cloning and Assembly Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
The complete kit for simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute or less, room temperature reaction
Invitrogen™ GeneArt™ Type IIs Assembly Kit Non-distribution product as customer accommodation.
Provides seamless cloning and assembly of up to 8 DNA fragments by simultaneous cleavage and ligation in a single reaction
Thermo Scientific™ RevertAid™ Premium Double Stranded cDNA Synthesis Kit
The Thermo Scientific Maxima H Minus Double-Stranded cDNA Synthesis Kit is a complete system for efficient synthesis of double-stranded cDNA from total RNA or mRNA.
Invitrogen™ GeneArt™ High-Order Genetic Assembly System Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
A highly efficient kit for the simultaneous and seamless assembly of up to 10 DNA fragments, totaling up to 110 Kbp in length, into any vector.
Invitrogen™ GeneArt™ pYES1L Vector with Sapphire™ Technology Non-distribution product as customer accommodation.
A ready-to-use, 9.3 Kb linearized BAC⁄YAC shuttle vector, used for the assembly of DNA fragments with the GeneArt™ High-Order Genetic Assembly System
Invitrogen™ GeneArt™ High-Order Vector Conversion Cassette Non-distribution product as customer accommodation.
A ready-to-use, 3.8kb linear BAC⁄YAC cassette, containing yeast replication and selection elements that can make any E.coli vector compatible with the GeneArt™ High-Order Genetic Assembly System
  spinner