RNA Isolation and Purification Products
MP Biomedicals™ Lysing Matrix D Available on GSA/VA Contract for Federal Government customers only.
FastPrep™ Lysing Matrix D are used primarily with FastPrep™ homogenizers or any other bead beater, for lysis of softer tissues like brain, liver, kidney, lung and spleen. Also used to lyse plant tissue such as leaves, roots, fruits as well as cultured cells and whole insects.
MP Biomedicals™ Lysing Matrix A, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
For all sample types except soil for the subsequent isolation of genomic DNA. MP Biomedicals Lysing Matrix A impact-resistant tubes contains garnet matrix and one 1/4 in. ceramic sphere.
MP Biomedicals™ Lysing Matrix B, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Contains 0.1mm silica spheres. Used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores. Lysing Matrix B can also be used as an additive for disruption by sonication.
MP Biomedicals™ Lysing Matrix E, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Lysing Matrix E is used primarily for environmental samples such as soil, sludge, wastwater and feces. Lysing Matrix E can also be effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumors and other difficult tissues.
MP Biomedicals™ Lysing Matrix S, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Use to break hard or brittle samples with minimal amounts of mechanical shearing. Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.
Promega™ Maxwell™ 16 LEV simplyRNA Blood Kit
Simple, automated RNA extraction
MP Biomedicals™ Lysing Matrix C, 2mL tubes Available on GSA/VA Contract for Federal Government customers only.
Contains 1mm silica spheres. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
MP Biomedicals™ Lysing Matrix A, 15mL tubes Available on GSA/VA Contract for Federal Government customers only.
For all sample types except soil for the subsequent isolation of genomic DNA. Each impact-resistant 15mL tube contains garnet matrix and ten 1/4 in. ceramic sphere. Lysing Matrix A is chemically inert and will not bind nucleic acids.
MP Biomedicals™ FastRNA™ Pro Green Kit Available on GSA/VA Contract for Federal Government customers only.
This kit is designed to efficiently isolate total RNA from any type of plant and animal tissue or cultured cells
MP Biomedicals™ FastRNA™ Pro Soil-Direct Kit Available on GSA/VA Contract for Federal Government customers only.
Used to isolate total cellular RNA from microorganisms and other specimens found in soil samples. Designed for use with FastPrep™ Instrument. Two levels of RNA purification, for different soil samples and downstream applications.
MP Biomedicals™ Lysing Matrix Y Available on GSA/VA Contract for Federal Government customers only.
For lysis of gram positive and negative bacteria, fungal tissue and spores, yeasts, molds, filamentous algaes and biofilms
MP Biomedicals™ Lysing Matrix H Available on GSA/VA Contract for Federal Government customers only.
Used to isolate RNA and Proteins from bacterial aggregates, biofilms, and whole insects
Thermo Scientific™ Buffers for GeneJET™ Blood RNA Kit
Purify high quality total RNA from fresh or frozen whole blood, as well as blood collected in PAXgene and Tempus Tubes
MP Biomedicals™ Lysing Matrix M Available on GSA/VA Contract for Federal Government customers only.
Tubes for optimal lysis used with FastPrep™ instruments
MP Biomedicals™ Geneclean™ SPIN Glassmilk™ Available on GSA/VA Contract for Federal Government customers only.
Used for Nucleic Acid isolation and purification
MP Biomedicals™ Lysing Matrix F Available on GSA/VA Contract for Federal Government customers only.
Used to isolate RNA and protein from samples. MP Briomedicals Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.
MP Biomedicals™ TallPrep™ Tubes
For use in Nucleic Acid Isolation and Purification
MP Biomedicals™ Lysing Matrix Z Available on GSA/VA Contract for Federal Government customers only.
Used primarily for lysis of animal and plant tissue, tougher stems, roots and seeds, and whole insectsDNase and RNase free.
MP Biomedicals™ Lysing Matrix C, 15mL tubes Available on GSA/VA Contract for Federal Government customers only.
Contains 1mm silica spheres. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
Promega™ QuantiFluor™ RNA System
Easily detects and quantifies small amounts of RNA in solution using a single-tube or plate reading fluorometer, supplied with QuantiFluor RNA Dye, Dilution Buffer and RNA standard
MP Biomedicals™ Lysing Matrix B, 50mL tubes Available on GSA/VA Contract for Federal Government customers only.
Contains 0.1mm silica spheres. Used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores. Lysing Matrix B can also be used as an additive for disruption by sonication.
MP Biomedicals™ Lysing Matrix A Available on GSA/VA Contract for Federal Government customers only.
For use in Nucleic Acid Isolation and Purification
MP Biomedicals™ Lysing Matrix E, 50mL tubes Available on GSA/VA Contract for Federal Government customers only.
Lysing Matrix E is used primarily for environmental samples such as soil, sludge, wastwater and feces. Lysing Matrix E can also be effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumors and other difficult tissues.
MP Biomedicals™ Lysing Matrix B, 96-well racks (1.2mL) Available on GSA/VA Contract for Federal Government customers only.
Contains 0.1mm silica spheres. Used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores. Lysing Matrix B can also be used as an additive for disruption by sonication.
MP Biomedicals™ FastRNA™ SPIN RNA Isolation Kit Available on GSA/VA Contract for Federal Government customers only.
Isolate total RNA from bacterial cell cultures and other microbial sources in approximately 15 minutes. Does not use phenol or reducing agents.
MP Biomedicals™ BigPrep™ Lysing Matrix Y Available on GSA/VA Contract for Federal Government customers only.
Used in Nucleic Acid Isolation and Purification. Ideal for cracking of very tough cell walls. DNase and RNase free.
MP Biomedicals™ Lysing Matrix K Available on GSA/VA Contract for Federal Government customers only.
Contains 0.8mm zirconium silicate beads
  spinner