UV Nucleic Acid Gel Stains
Invitrogen™ UltraPure™ 10mg/mL Ethidium Bromide Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Sensitive fluorescent dye detects nucleic acids in agarose gels and cesium chloride gradients
Lonza™ GelStar™ Nucleic Acid Gel Stain
Use to detect small quantities of DNA and RNA.  GelStar™ Nucleic Acid Gel Stain exhibits exceptional signal-to-noise ratio with minimal background. 
MP Biomedicals™ BBXG (With Glycerol)
Used as a loading dye for DNA samples. MP Biomedicals BBXG (With Glycerol) is a 10x solution of Bromophenol Blue, Xylene Cyanol FF and Glycerol that is used as a loading buffer for DNA gel electrophoresis.
  spinner