Blocking Buffers for Protein Methods
Thermo Scientific™ Blocker™ Casein Blocking Buffers Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and other samples with these ready-to-use PBS or TBS solutions of purified casein for Western blot, ELISA and other detection methods.
Thermo Scientific™ Blocker™ BSA Blocking Buffers Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and other samples with these ready-to-use 10X PBS or TBS solutions of bovine serum albumin for Western blot, ELISA and other detection methods.
Thermo Scientific™ Blocker™ BLOTTO Blocking Buffer Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and other samples with this ready-to-use 5% solution of nonfat powdered milk in TBS for Western blot, ELISA and other detection methods.
Thermo Scientific™ Pierce™ Protein-Free Blocking Buffers Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and other samples with these ready-to-use PBS and TBS formulations of a non-protein compound to yield extremely low background.
Thermo Scientific™ Pierce™ Clear Milk Blocking Buffer
Thermo Scientific™ Pierce Clear Milk Blocking Buffer is a long shelf-life, high performance replacement for homemade milk blocking buffers in Western blotting applications.
Thermo Scientific™ Pierce™ Fast Blocking and Wash Buffers
Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with this ready-to-use blocker solution.
EMD Millipore™ Immunoblot Blocking Reagent
For use in western blots as a blocking agent
  spinner