Buffers and Diluents
Thermo Scientific™ SuperBlock™ Blocking Buffers Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and tissue samples with these optimized solutions containing a protein that provides fast and effective blocking for Western blot, ELISA and IHC.
Invitrogen™ Novex™ NuPAGE™ MES SDS Running Buffer (20X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Made with high-purity reagents and are strictly quality controlled
BD Brilliant Stain Buffer
Designed to complement multicolor flow cytometry experiments. Brilliant Stain Buffer is a buffer for the immunofluorescent staining of cells.
Invitrogen™ Novex™ NuPAGE™ MOPS SDS Running Buffer (20X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Made with high-purity reagents and are strictly quality controlled
Thermo Scientific™ Pierce™ StartingBlock™ Blocking Buffers Available on GSA/VA Contract for Federal Government customers only.
Block membranes, plates and other samples with these ready-to-use PBS or TBS formulations of a special protein blocker for exceptional nonspecific blocking and broad compatibility.
Invitrogen™ Novex™ NuPAGE™ Transfer Buffer (20X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Use to transfer proteins from NuPAGE Novex gels to membranes for Western blotting
R&D Systems™ Reagent Diluent Concentrate 2, 10X Available on GSA/VA Contract for Federal Government customers only.
10X Concentrated 10% BSA Solution for DuoSets
Thermo Scientific™ Pierce™ Guardian™ Peroxidase Conjugate Stabilizer/Diluent
Preserve diluted (1-1,000ng/mL) HRP conjugates for more than six months at room temperature without significant loss of activity using Guardian Peroxidase Conjugate Stabilizer/Diluent.
Invitrogen™ Novex™ NuPAGE™ Sample Reducing Agent (10X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Ready-to-use reagent to reduce protein samples for protein gel electrophoresis
Thermo Scientific™ 6X DNA Loading Dye Available on GSA/VA Contract for Federal Government customers only.
Prepare DNA markers and samples for loading on agarose or polyacrylamide gels with Thermo Scientific™ 6X DNA Loading Dye.
R&D Systems™ Stop Solution 2N Sulfuric Acid Available on GSA/VA Contract for Federal Government customers only.
For stopping the action of HRP on the substrate, tetramethylbenzidine
Invitrogen™ Novex™ NuPAGE™ Antioxidant Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Proprietary reagent that maintains proteins in a reduced state during protein gel electrophoresis
GE Healthcare Pharmalyte™ Carrier Ampholytes Available on GSA/VA Contract for Federal Government customers only.
Use in Isoelectric focusing. GE Healthcare Pharmalyte™ Carrier Ampholytes prepared by the co-polymerization of glycine, glycylglycine, amines, and epichlorohydrin, are available in five broad-range and four narrow-range pH intervals.
Invitrogen™ Novex™ NuPAGE™ Tris-Acetate SDS Running Buffer (20X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Formulated for separation of proteins, in their denatured state, on NuPAGE Novex Tris-Acetate gels
R&D Systems™ Quantikine Wash Buffer 1 Available on GSA/VA Contract for Federal Government customers only.
Concentrate that can be diluted to a working concentration and used for the complete removal of any unbound sample or conjugate in the wells of an ELISA
Invitrogen™ Novex™ Tris-Glycine Transfer Buffer (25X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Made with high-purity reagents and are strictly quality controlled
Invitrogen™ Novex™ NativePAGE™ Running Buffer (20X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used to make the NativePAGE Cathode and Anode Running Buffers
Thermo Scientific™ Pierce™ SEA BLOCK™ Blocking Buffer Available on GSA/VA Contract for Federal Government customers only.
Block mammalian tissue samples for immunohistochemistry (IHC) and samples for other methods with steelhead salmon serum in PBS.
Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Formulated for separation of proteins, in their denatured state, on Novex Tris-Glycine gels
Invitrogen™ Novex™ Bolt™ Transfer Buffer (20X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Optimized for the transfer of proteins from Bolt Bis-Tris Plus gels to membranes for western blotting
Invitrogen™ Novex™ NativePAGE™ Running Buffer Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Contains the NativePAGE Running Buffer (20X)
Invitrogen™ Novex™ NativePAGE™ Sample Prep Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Includes sample preparation reagents for native gel electrophoresis
BD Perm / Wash™ Buffer I (10x) Available on GSA/VA Contract for Federal Government customers only.
Intracellular staining of post-translationally modified signaling proteins
Invitrogen™ Novex™ IEF Cathode Buffer pH 3-10 (10X) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
An optimized pre-mixed IEF cathod buffer
Thermo Scientific™ Pierce™ ELISA Blocker Blocking Buffer
Increases signal-to-noise ratio and minimizes background
Thermo Scientific™ Pierce™ ELISA Wash Buffers
Concentrated wash buffer for efficient use of storage in the lab
  spinner