Goods Buffers
MES monohydrate, 99+%, for biochemistry, ACROS Organics™
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N Synonym: 2-(N-Morpholino)ethanesulfonic acid monohydrate PubChem CID: 16218417 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid;hydrate SMILES: C1COCCN1CCS(=O)(=O)O.O
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N Synonym: N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
MES (Fine White Crystals), Fisher BioReagents
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N Synonym: 2-(4-Morpholino)ethane Sulfonic Acid, 2-(N-Morpholino)ethanesulfonic acid hydrate PubChem CID: 16218417
HEPES, 99%, for biochemistry, ACROS Organics™
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N Synonym: 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
PIPES, 98.5+%, for biochemistry, ACROS Organics™
CAS: 5625-37-6 Molecular Formula: C8H18N2O6S2 Molecular Weight (g/mol): 302.36 InChI Key: IHPYMWDTONKSCO-UHFFFAOYSA-N Synonym: 1, 4-Piperazinebis(ethanesulfonic acid) PubChem CID: 79723 ChEBI: CHEBI:44933 IUPAC Name: 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
Alfa Aesar™ HEPES, 1.0M buffer soln., pH 7.0
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
HEPES Sodium Salt (White Powder), Fisher BioReagents
CAS: 75277-39-3 Molecular Formula: C8H17N2NaO4S Molecular Weight (g/mol): 260.284 InChI Key: RDZTWEVXRGYCFV-UHFFFAOYSA-M PubChem CID: 2724248 ChEBI: CHEBI:46758 IUPAC Name: sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate SMILES: C1CN(CCN1CCO)CCS(=O)(=O)[O-].[Na+]
ACES, 99+%, for biochemistry, ACROS Organics™
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N Synonym: N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N
MES Monohydrate, 98%, Alfa Aesar™
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid;hydrate SMILES: C1COCCN1CCS(=O)(=O)O.O
HEPES, ≥99% (Dry Basis), Ultrapure, Affymetrix/USB™
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N Synonym: N-[2-Hydroxyethyl]piperazine-N'-[2-Ethanesulfonic acid] PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
MES, 99+%, <1% water, ACROS Organics™
CAS: 4432-31-9 Molecular Formula: C6H13NO4S Molecular Weight (g/mol): 195.233 MDL Number: MFCD00006181 InChI Key: SXGZJKUKBWWHRA-UHFFFAOYSA-N Synonym: 2-(N-Morpholino)ethanesulfonic acid PubChem CID: 78165 ChEBI: CHEBI:39005 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid SMILES: C1COCCN1CCS(=O)(=O)O
TES (White Crystals), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
CAS: 7365-44-8 Molecular Formula: C6H15NO6S Molecular Weight (g/mol): 229.247 InChI Key: JOCBASBOOFNAJA-UHFFFAOYSA-N Synonym: N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid PubChem CID: 81831 ChEBI: CHEBI:44356
HEPES Buffer, 1M Solution, Cell Culture Grade, MP Biomedicals
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N PubChem CID: 23831 ChEBI: CHEBI:42334
Alfa Aesar™ HEPES, 1.0M buffer soln., pH 7.5
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
MES hydrate, 99+%, Acros Organics™
CAS: 1266615-59-1 Molecular Formula: C6H13NO4S·xH2O MDL Number: MFCD00149409 Synonym: 4-Morpholinoethanesulfonic acid hydrate, MES
HEPES, Free Acid, Ultrapure Bioreagent, J.T.Baker™
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
Alfa Aesar™ MES, 1.0M buffer soln., pH 6.0
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 MDL Number: MFCD00283848 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid;hydrate SMILES: C1COCCN1CCS(=O)(=O)O.O
Tricine (White Crystals), Fisher BioReagents
CAS: 5704-04-1 Molecular Formula: C6H13NO5 Molecular Weight (g/mol): 179.17 InChI Key: SEQKRHFRPICQDD-UHFFFAOYSA-N Synonym: N-[Tris-(hydroxymethyl)methyl]glycine PubChem CID: 79784 ChEBI: CHEBI:39063
Tricine, 98%, for biochemistry, ACROS Organics™
CAS: 5704-04-1 Molecular Formula: C6H13NO5 Molecular Weight (g/mol): 179.17 MDL Number: MFCD00004277 InChI Key: SEQKRHFRPICQDD-UHFFFAOYSA-N Synonym: N-[Tris(hydroxymethyl)methyl]glycine, N-(2-Hydroxy-1, 1-bis(hydroxymethyl)ethyl)glycine PubChem CID: 79784 ChEBI: CHEBI:39063 IUPAC Name: 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid SMILES: C(C(=O)O)NC(CO)(CO)CO
TES, 99%, for biochemistry, ACROS Organics™
CAS: 7365-44-8 Molecular Formula: C6H15NO6S Molecular Weight (g/mol): 229.247 MDL Number: MFCD00007532 InChI Key: JOCBASBOOFNAJA-UHFFFAOYSA-N Synonym: 2-[Tris(hydroxymethyl)methylamino]-1-ethanesulfonic acid PubChem CID: 81831 ChEBI: CHEBI:44356 IUPAC Name: 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NC(CO)(CO)CO
HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free, ACROS Organics™
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N Synonym: 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid, 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure, Affymetrix/USB™
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
PIPES, disodium salt, 99%, ACROS Organics™
CAS: 76836-02-7 Molecular Formula: C8H16N2Na2O6S2 Molecular Weight (g/mol): 346.324 InChI Key: GMHSTJRPSVFLMT-UHFFFAOYSA-L Synonym: 1, 4-Piperazinediethanesulfonic acid disodium salt PubChem CID: 173553 ChEBI: CHEBI:63055 IUPAC Name: disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate SMILES: C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+]
Tricine, ≥98.5% (Dry Basis), Ultrapure, Affymetrix/USB™
CAS: 5704-04-1 Molecular Formula: C6H13NO5 Molecular Weight (g/mol): 179.17 MDL Number: MFCD00004277 InChI Key: SEQKRHFRPICQDD-UHFFFAOYSA-N Synonym: N-Tris(Hydroxymethyl)methylglycine PubChem CID: 79784 ChEBI: CHEBI:39063 IUPAC Name: 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid SMILES: C(C(=O)O)NC(CO)(CO)CO
Alfa Aesar™ HEPES, 1.0M buffer soln., pH 8.0
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
Alfa Aesar™ MES, 1.0M buffer soln., pH 5.0
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 MDL Number: MFCD00283848 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid;hydrate SMILES: C1COCCN1CCS(=O)(=O)O.O
Alfa Aesar™ MES, 1.0M buffer soln., pH 6.5
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 MDL Number: MFCD00283848 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid;hydrate SMILES: C1COCCN1CCS(=O)(=O)O.O
Alfa Aesar™ HEPES-buffered saline, pH 7.0 (2X for transfection)
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
PIPES, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
CAS: 5625-37-6 Molecular Formula: C8H18N2O6S2 Molecular Weight (g/mol): 302.36 MDL Number: MFCD00006159 InChI Key: IHPYMWDTONKSCO-UHFFFAOYSA-N Synonym: Piperazine-N, N'-bis[2-ethanesulfonic acid] PubChem CID: 79723 ChEBI: CHEBI:44933 IUPAC Name: 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
  spinner