Membrane Stripping Buffers
Thermo Scientific™ Restore™ Plus Western Blot Stripping Buffer Available on GSA/VA Contract for Federal Government customers only.
Remove high-affinity antibodies from membranes so they can be reprobed and detected with chemiluminescent substrates.
Thermo Scientific™ Restore™ Western Blot Stripping Buffer Available on GSA/VA Contract for Federal Government customers only.
Safely and effectively remove primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
EMD Millipore™ ReBlot Plus Kit and Reagents Available on GSA/VA Contract for Federal Government customers only.
Blots can be stripped of antibodies in approximately 15 minutes at room temperature
Thermo Scientific™ Restore™ Fluorescent Western Blot Stripping Buffer
Quickly and gently remove primary and near-infrared (IR) dye-labeled secondary antibodies from Western blots.
  spinner