Nucleic Acid EPP Loading Buffers
Promega™ Blue/Orange Loading Dye, 6X
For loading DNA samples onto nondenaturing gels
Lonza™ Formaldehyde Sample Buffer
Ideal when samples are to be recovered from agarose gels
Promega™ Labeling 5X Buffer
Contains random synthetic hexadeoxynucleotide primers for random-primed labeling of linear template DNA with radionucleotides
EMD Millipore™ Novagen™ 6X DNA Gel Loading Buffer Available on GSA/VA Contract for Federal Government customers only.
Intended for gel electrophoresis of DNA fragments or PCR products.
Lonza™ Ficoll™ DNA Loading Buffer, 6X
Ideal for improving resolution and separation of DNA fragments
EMD Millipore™ Novagen™ 6X Loading Buffer Available on GSA/VA Contract for Federal Government customers only.
Intended for gel electrophoresis of DNA fragments or PCR products.
Promega™ 5X Colorless GoTaq™ Flexi Buffer
Samples can be loaded directly onto gels without the need for loading dye
Promega™ Block and Sample 5X Buffer
Optimized for use in the Emax™ ImmunoAssay Systems for BDNF, GDNF, NGF and NT-3
Affymetrix/USB™ RapidRun™ Ultrapure Agarose Buffer
For high voltage, fast agarose gel electrophoresis (20-30 volts/cm). Affymetrix/USB™ RapidRun™ Ultrapure Agarose Buffer is compatible with commercial gel extraction kits by adjusting pH to 5 with sodium acetate.
  spinner