Antibiotics, Buffers and Supplements
MES monohydrate, 99+%, for biochemistry, ACROS Organics™
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N Synonym: 2-(N-Morpholino)ethanesulfonic acid monohydrate PubChem CID: 16218417 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid;hydrate SMILES: C1COCCN1CCS(=O)(=O)O.O
HEPES sodium salt, 99%, ACROS Organics™
CAS: 75277-39-3 Molecular Formula: C8H17N2NaO4S Molecular Weight (g/mol): 260.284 MDL Number: MFCD00036463 InChI Key: RDZTWEVXRGYCFV-UHFFFAOYSA-M Synonym: 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt PubChem CID: 2724248 ChEBI: CHEBI:46758 IUPAC Name: sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate SMILES: C1CN(CCN1CCO)CCS(=O)(=O)[O-].[Na+]
MES Monohydrate, 98%, Alfa Aesar™
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 MDL Number: MFCD00149409 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N PubChem CID: 16218417 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid;hydrate SMILES: C1COCCN1CCS(=O)(=O)O.O
HEPES, 99%, for biochemistry, ACROS Organics™
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 MDL Number: MFCD00006158 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N Synonym: 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
Gibco™ Trypsin-EDTA (0.25%), Phenol red Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Made from trypsin powder, irradiated mixture of proteases derived from porcine pancreas
Corning™ Matrigel™ GFR Membrane Matrix Available on GSA/VA Contract for Federal Government customers only.
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ GFR Basement Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
Corning™ Cell Culture Phosphate Buffered Saline (1X) Available on GSA/VA Contract for Federal Government customers only.
With low endotoxin content. Without calcium and magnesium.
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents
CAS: 7365-45-9 Molecular Formula: C8H18N2O4S Molecular Weight (g/mol): 238.302 InChI Key: JKMHFZQWWAIEOD-UHFFFAOYSA-N Synonym: N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid PubChem CID: 23831 ChEBI: CHEBI:42334 IUPAC Name: 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid SMILES: C1CN(CCN1CCO)CCS(=O)(=O)O
Gibco™ MarrowMAX™ Bone Marrow Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Fully-supplemented medium for short-term culture of bone marrow and other hematopoietic cells for cytogenetic studies and in vitro diagnostic procedures
Corning™ Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, LDEV-Free
Suited for applications requiring highly defined basement membrane preparation.
Bovine Pituitary Extract, BT-215, Alfa Aesar™
Extract prepared from bovine pituitary glands.
Corning™ Matrigel™ Membrane Matrix Available on GSA/VA Contract for Federal Government customers only.
Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ Membrane Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.
Gibco™ TrypLE Express Enzyme (1X), No Phenol Red Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Unlike porcine trypsin, free of animal-derived components
Corning™ Epidermal Growth Factor (EGF), Mouse Natural (Receptor Grade)
Enhance the proliferation of a wide variety of cell types
Corning™ 0.25% Trypsin, 0.1% EDTA in HBSS w/o Calcium, Magnesium and Sodium Bicarbonate Available on GSA/VA Contract for Federal Government customers only.
Used to enzymatically release adherent cells from tissue culture plates for passaging.
Corning™ Matrigel™ hESC-Qualified Matrix Available on GSA/VA Contract for Federal Government customers only.
Compatible with mTeSR™1-devloped by StemCell Technologies. Corning™ Matrigel™ hESC-Qualified Matrix is widely accepted as an alternative substrate for the culture of hES cells as well as human induced pluripotent stem (iPS) cells.
ACES, 99+%, for biochemistry, ACROS Organics™
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N Synonym: N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N
Corning™ DMSO (Dimethyl Sulfoxide) Available on GSA/VA Contract for Federal Government customers only.
For cryopreservation of cells along with glycerol, to prevent damage to cell membrane during freezing
MP Biomedicals™ Mycoplasma Removal Agent
Eliminates mycoplasma contamination from cell cultures
BD Bacto™ Dehydrated Culture Media Additive: Yeast Extract
Formulation of culture media for cultivation of heterotrophic microorganisms.  BD Difco™ Bacto™ Dehydrated Culture Media Additive: Yeast Extract is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
Gibco™ Trypsin-EDTA (0.05%), Phenol red Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas
Gibco™ GlutaMAX™ Supplement Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
An alternative to L-glutamine, with increased stability that improves cell health
MES (Fine White Crystals), Fisher BioReagents
CAS: 145224-94-8 Molecular Formula: C6H15NO5S Molecular Weight (g/mol): 213.248 InChI Key: MIIIXQJBDGSIKL-UHFFFAOYSA-N Synonym: 2-(4-Morpholino)ethane Sulfonic Acid, 2-(N-Morpholino)ethanesulfonic acid hydrate PubChem CID: 16218417 IUPAC Name: 2-morpholin-4-ylethanesulfonic acid;hydrate SMILES: C1COCCN1CCS(=O)(=O)O.O
Corning™ Endothelial Cell Growth Supplement (ECGS) Available on GSA/VA Contract for Federal Government customers only.
Ideal for endothelial cell culture
  spinner