Amino Acids
Promega™ Amino Acid Mixtures
Compatible for use in the Flexi™ Lysate, TNT™ Lysate and standard Rabbit Reticulocyte Lysate Systems as well as in the Wheat Germ Extract and E. coli S30 Systems
Lonza™ Ultraglutamine I Supplement
Substitute for L-glutamine on an equimolar basis for most adherent and suspension cultures.  Lonza™ Ultraglutamine I Supplement avoids accumulation of toxic ammonia produced by spontaneous degradation in the medium.
MilliporeSigma™ EmbryoMax™ MEM Non Essential Amino Acids
available in a 100mL format and may be used for routine mouse embryonic stem cell culture applications.
  spinner