Nutritional Supplements
Corning™ Cell Culture Phosphate Buffered Saline (1X) Available on GSA/VA Contract for Federal Government customers only.
With low endotoxin content. Without calcium and magnesium.
Corning™ HEPES, Liquid Available on GSA/VA Contract for Federal Government customers only.
An alternative to use of bicarbonate for buffering
BD Bacto™ Dehydrated Culture Media Additive: Yeast Extract
Formulation of culture media for cultivation of heterotrophic microorganisms.  BD Difco™ Bacto™ Dehydrated Culture Media Additive: Yeast Extract is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
Corning™ DMSO (Dimethyl Sulfoxide) Available on GSA/VA Contract for Federal Government customers only.
For cryopreservation of cells along with glycerol, to prevent damage to cell membrane during freezing
Corning™ ITS Premix Universal Culture Supplement Available on GSA/VA Contract for Federal Government customers only.
Stimulates proliferation of a variety of cells under serum-reduced conditions. Corning™ ITS Premix Universal Culture Supplement contains insulin, human transferrin and selenous acid, the three most universally esential components of defined culture media.
BD Bacto™ Dehydrated Culture Media Additive: Tryptone Available on GSA/VA Contract for Federal Government customers only.
Enzymatic digest of casein. BD Difco™ Bacto™ Dehydrated Culture Media Additive: Tryptone is part of a large selection of dehydrated media.
Corning™ Tris Base Buffer, Powder
Used in buffer solutions in biochemical and molecular biological research
Corning™ Endothelial Cell Growth Supplement (ECGS) Available on GSA/VA Contract for Federal Government customers only.
Ideal for endothelial cell culture
BD Bacto™ Animal-Origin Media Supplement: Tryptone
For use with dehydrated media culture
Corning™ L-glutamine Solution Available on GSA/VA Contract for Federal Government customers only.
Essential amino acid and a key component of culture media
BD Bacto™ Proteose Peptone Available on GSA/VA Contract for Federal Government customers only.
Used in preparing microbiological culture media and in producing bacterial toxins. BD™ Bacto™ Proteose Peptone is a highly nutritious, animal-based media. The enzymatic digest of protein is high in proteoses and provides nitrogen in a form that is readily available for bacterial growth.
MilliporeSigma™ GS System 100mM L-Methionine sulfoximine
utilizes specific vectors for cell culture transfection.
Lonza™ BioWhittaker™ Water for Cell Culture
CAS: 7732-18-5 Molecular Formula: H2O Molecular Weight (g/mol): 18.015 InChI Key: XLYOFNOQVPJJNP-UHFFFAOYSA-N Synonym: dihydrogen oxide, dihydrogen monoxide PubChem CID: 962 ChEBI: CHEBI:15377 IUPAC Name: oxidane SMILES: O
MilliporeSigma™ Chemicon™ ESGRO™ Leukemia Inhibitory Factor (LIF), 10 million units/1mL Available on GSA/VA Contract for Federal Government customers only.
Optimized formulation of recombinant mouse LIF (mLIF) for in vitro maintenance of mouse ES and iPS cells.
BD Difco™ Dehydrated Culture Media: TC Yeastolate UF
Used for preparing bacterial, insect and mammalian cell culture media.
BD BBL™ Dehydrated Culture Media Additive: Yeast Extract
Water-soluble extract of autolyzed yeast cells suitable for use in culture media
Corning™ MEM Vitamin Solution Available on GSA/VA Contract for Federal Government customers only.
Addition to medium improves growth and viability of propagating cells in culture while reducing the biosynthetic burden on these cells
BD Bacto™ Dehydrated Culture Media Additive: Bottle Yeast Extract
Used with dehydrated culture media.   BD Difco™ Bacto™ Dehydrated Culture Media Additive: Bottle Yeast Extract is an important additive.
Corning™ Trypan Blue Available on GSA/VA Contract for Federal Government customers only.
Can be used for variety cell counting methods requiring information on cell count and viability.
  spinner