Single Chamber Flat Sided Cell Culture Flasks
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Corning™ Cell Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polstyrene for optimum visibility and distortion-free microscopic viewing. Corning™ Cell Culture Treated Flasks are available in a variety of sizes, designs and cap styles to meet laboratory diagnostic needs.
Falcon™ Tissue Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically clear virgin polystyrene for easy viewing of sample or cells. Corning™ Falcon™ Tissue Culture Treated Flasks features vacuum-gas plasma tissue culture treatment to ensure consistent growth surface.
Thermo Scientific™ Nunc™ EasYFlask™ Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Enhance cell attachment, growth and differentiation with Thermo Scientific™ Nunc™ Cell Culture Treated EasYFlasks™.
Fisherbrand™ Surface Treated Sterile Tissue Culture Flasks, Vented Cap
Perfect for cell growth, cell yields aim on little or medium input volume.
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Thermo Scientific™ BioLite Cell Culture Treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Perform general cell culture procedures more economically with Thermo Scientific™ BioLite Cell Culture Treated Flasks, guaranteed to be sterile, non-pyrogenic and non-cytotoxic.
Corning™ U-Shaped Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Designed to improve usability while maintaining the same environment for cell growth as previous designs. Corning™ U-Shaped Cell Culture Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Greiner Bio-One CELLSTAR™ TC Treated Cell Culture Flasks
Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR™ TC Treated Cell Culture Flasks are free of detectable DNase, RNase and human DNA.
Corning™ Cell Culture Untreated Flasks Available on GSA/VA Contract for Federal Government customers only.
Printed with lot numbers for ease in traceability
Corning™ Ultra-Low Attachment Flasks Available on GSA/VA Contract for Federal Government customers only.
Features a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation. Corning™ Ultra-Low Attachment Flasks have an ergonomic shape that reduces the number of corners, improves cell scraping and allows the use of a larger pipet.
Corning™ CellBIND™ Surface Cell Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.
Greiner Bio-One CELLSTAR™ TC Treated Lo-Profile Cell Culture Flasks
Improved cell adhesion through physical surface treatment. Greiner Bio-One CELLSTAR™ TC Treated Lo-Profile Cell Culture Flasks is free of detectable DNase, RNase and human DNA.
Thermo Scientific™ Nunc™ Non-treated Flasks Available on GSA/VA Contract for Federal Government customers only.
Minimize the chance of contamination with these high-quality, optically clear polystyrene Thermo Scientific™ Nunc™ Non-treated Flasks for suspension cell cultures.
Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap Available on GSA/VA Contract for Federal Government customers only.
Pre-coated with Collagen I for improved cell attachment and an increased proliferation rate of transfected mammalian cells, primary cells and stem cells. Corning™ BioCoat™ Collagen I Rectangular Canted Neck Cell Culture Flask with Vented Cap provides highly consistent and biologically functional precoated surfaces to more closely mimic in vivo environments for cell culture applications.
Greiner Bio-One CELLCOAT™ Collagen Type I Coated Cell Culture Flask
Ideal for cell cultivation in serum-free or serum-reduced medium
Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps Available on GSA/VA Contract for Federal Government customers only.
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps.
Corning™ Cell Culture Flask with Bar Code
Features nonrepeating bar code for use in automation
Corning™ Primaria™ Tissue Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
Falcon™ Nontreated Tissue Culture Flasks Available on GSA/VA Contract for Federal Government customers only.
Similar to the surface of a bacteriological Petri dish
Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask Available on GSA/VA Contract for Federal Government customers only.
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask is ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
Greiner Bio-One CELLSTAR™ Suspension Culture Flask with Filter Cap
Ideal for suspension cultures, hybridoma and embryonic stem cells. Greiner Bio-One CELLSTAR™ Suspension Culture Flask with Filter Cap is free of detectable DNase, RNase and human DNA.
Thermo Scientific™ Nunclon™ Sphera™ Flasks
Features a superior cell culture surface with demonstrated performance that elevates stem cell and cell culture research.
Corning™ BioCoat™ Fibronectin Rectangular Canted Neck Culture Flask
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
Corning™ PureCoat™ Carboxyl Rectangular Canted Neck Cell Culture Flasks with Vented Cap
Negatively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays
Corning™ BioCoat™ Laminin Rectangular Canted Neck Cell Culture Flask w/Plug Seal Cap
Tested for their ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells
Corning™ PureCoat™ Fibronectin Peptide Cell Culture Flasks with Vented Cap
Coated with biologically active synthetic animal-free peptides that are covalently linked to a proprietary surface
Argos Technologies™ CELLine Bioreactor Flasks
Specialized cell culture flask for production of monoclonal antibodies or expressed proteins from suspension cells
Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks Available on GSA/VA Contract for Federal Government customers only.
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  spinner