Insect Cell Culture Media
Corning™ Insectagro™ Sf9 Serum-free/Protein-free Medium, 1X, With L-glutamine Available on GSA/VA Contract for Federal Government customers only.
Formulated to support propagation of Sf9 insect cells in culture.
Corning™ Poloxamer 188, Powder Available on GSA/VA Contract for Federal Government customers only.
Acts as surfactant used in cell culture to protect cells in suspension against possible damage during transfer, freezing and thawing, and stirring.
Corning™ Poloxamer 188, 10% Solution Available on GSA/VA Contract for Federal Government customers only.
Acts as surfactant used in cell culture to protect cells in suspension against possible damage during transfer, freezing and thawing, and stirring.
Lonza™ BioWhittaker Insect-XPRESS Insect Cell Culture Media (1X) with L-Glutamine
Designed to support the growth of insect cell lines derived from Spodoptera frugiperda (Sf9 and Sf21). BioWhittaker™ Insect-XPRESS™ Insect Cell Culture Media (1X) with L-Glutamine supports superior production of recombinant proteins when using the Baculovirus Expression Vector System (BEVS).
Corning™ Insectagro™ DS2 Serum-Free/Protein-Free Medium, 1X, without L-glutamine Available on GSA/VA Contract for Federal Government customers only.
Used for growth and maintenance of Drosophila Schneider 2 (DS2) cells intended for use in heterologous protein expression.
Corning™ Hink's TNM-FH Medium Available on GSA/VA Contract for Federal Government customers only.
Used for low endotoxin content and better results.
Corning™ Grace's Insect Basal medium (Vaughn Mod.) Available on GSA/VA Contract for Federal Government customers only.
Used as basal medium for growth and maintenance of cell lines derived from lepidopterans.
Lonza™ BioWhittaker™ Insect Cell Culture Media: Schneider's Drosophila Medium (Modified)
formulated for the growth of S2 cells from the fruit fly, Drosophila melanogaser. BioWhittaker™ Insect Cell Culture Media: Schneider's Drosophila Medium (Modified) may be used to culture a variety of other insect cell lines as well.
EMD Millipore Novagen™ TriEx™ Insect Cell Medium Available on GSA/VA Contract for Federal Government customers only.
Serum-free medium optimized for growth and protein production of TriEx Sf9 Cells
Mirus Bio™ Poloxamer 188, 10% Solution
Added to batch mammalian cell cultures to reduce foaming
Lonza™ BioWhittaker™ Insect Cell Culture Media: Grace's Insect Medium
Chemically-defined media optimized for specific cell types
HyClone™ SFX-Insect™ Cell Culture Media
Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.
GE Healthcare HyClone™ SFX Insect™ Cell Culture Media PROMO
Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.
  spinner