Mamallian Cell Culture Media
Gibco™ Recovery™ Cell Culture Freezing Medium, DMSO (10%)
Complete, ready-to-use freezing medium for cryopreservation of a wide variety of mammalian cells, including CHO-S, CHO-K1, HEK 293, Jurkat, and NIH 3T3
Corning™ Minimum Essential Medium Eagle (Mod.)
Formulated without sodium bicarbonate to prevent clumping due to moisture absorption
Gibco™ Expi293™ Expression Medium
Designed for growth, transfection of suspension-adapted human embryonic kidney (HEK) 293 cells
Gibco™ DMEM, Powder, High Glucose, No Sodium Bicarbonate Buffer
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, High Glucose, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Corning™ DMEM with L-Glutamine, 4.5g/L Glucose and Sodium Pyruvate 
Subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ DMEM with L-Glutamine, 4.5g/L Glucose and Sodium Pyruvate is hybridoma tested.
Gibco™ DMEM, High Glucose
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose is used in cell culture processing applications.
Gibco™ RPMI 1640 Medium
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM, Powder, High Glucose, Pyruvate, No Sodium Bicarbonate Buffer
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, High Glucose, Pyruvate, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Gibco™ Neurobasal™ Medium
Used in the growth of neuronal cells from hippocampus, cortex and otregions of the brain. Gibco™ Neurobasal™ Medium is a basal medium that meets the special cell culture requirements of pre-natal and embryonic neuronal cells when used with Gibco™ B-27™ Supplement.
Gibco™ DMEM/F-12, Powder
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, Powder is used in cell culture processing applications.
Gibco™ DMEM, High Glucose, Pyruvate
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, Pyruvate is used in cell culture processing applications.
Gibco™ Essential 8™ Flex Medium Kit
Essential 8™ Flex is a xeno-free and feeder-free medium that supports the culture of pluripotent stem cells (PSCs) without the need for weekend feeding. Gibco™ Essential 8™ Flex Medium Kit is provided in a convenient two-component kit (500mL basal medium and 10mL supplement).
Gibco™ Opti-MEM™ I Reduced Serum Medium
An improved Minimal Essential Medium that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology
HyClone™ HyQ™-RS Reduced Serum Media
For applications requires lower concentration of serum supplementation
Corning™ RPMI 1640 Medium (Mod.) 1X with L-Glutamine 
Guaranteed low endotoxin levels ≤0.025EU/mL
Gibco™ DMEM/F-12, Powder, HEPES
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, Powder, HEPES is used in cell culture processing applications.
GE Healthcare HyClone™ CDM4PERMAb cell culture media
Chemically defined Animal-Derived Component-Free (ADCF) media for use with PER.C6 cells
Corning™ Dulbecco's Modification of Eagle's Medium (DMEM) 1X1, Powder 
Subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ Dulbecco's Modification of Eagle's Medium (DMEM) 1X1, Powder is hybridoma tested.
Gibco™ Advanced DMEM/F-12
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ Advanced DMEMF/12 is used in cell culture processing applications.
Gibco™ RPMI 1640 Medium (ATCC Modification), HEPES, Liquid
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM/F-12, HEPES
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, HEPES is used in cell culture processing applications.
Corning™ NutriStem™ hPSC XF Medium
Supports long-term culture of human pluripotent stem cells
Corning™ DMEM With L-Glutamine and 4.5g/L Glucose; Without Sodium Pyruvate 
Subsititution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation. Corning™ DMEM With L-Glutamine and 4.5g/L Glucose; Without Sodium Pyruvate is hybridoma tested.
HyClone™ ADCF-MAb Media
Increase process yields for the manufacture of monoclonal antibodies (MAbs) with HyClone™ ADCF-MAb Media, a protein-free media containing no animal-derived components.
HyClone™ SFM4Transfx-293 Media
Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.
Lonza™ BioWhittaker™ Hybridoma Serum-Free Culture Media: HL-1 Complete
Prescreened in actual hybridoma systems to ensure minimal variation