Cell Culture Media
Gibco™ DMEM, Powder, High Glucose, No Sodium Bicarbonate Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, High Glucose, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Gibco™ DMEM, High Glucose Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose is used in cell culture processing applications.
Gibco™ Expi293™ Expression Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed for growth, transfection of suspension-adapted human embryonic kidney (HEK) 293 cells
Gibco™ RPMI 1640 Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM, High Glucose, Pyruvate Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, Pyruvate is used in cell culture processing applications.
Gibco™ DMEM/F-12, Powder Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, Powder is used in cell culture processing applications.
Gibco™ DMEM, Powder, High Glucose, Pyruvate, No Sodium Bicarbonate Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, High Glucose, Pyruvate, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Gibco™ Opti-MEM™ I Reduced Serum Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
An improved Minimal Essential Medium that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology
Gibco™ Advanced DMEM/F-12 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ Advanced DMEMF/12 is used in cell culture processing applications.
Gibco™ DMEM/F-12, HEPES Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, HEPES is used in cell culture processing applications.
Gibco™ DMEM, High Glucose, GlutaMAX™ Supplement, Pyruvate Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, GlutaMAX™ Supplement, Pyruvate is used in human ex vivo tissue and cell culture processing applications.
Gibco™ DMEM/F-12, GlutaMAX™ Supplement Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, GlutaMAX™ Supplement is used in cell culture processing applications.
Gibco™ RPMI 1640 Medium (ATCC Modification), HEPES, Liquid Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM/F-12 Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM/F-12 is used in cell culture processing applications.
Gibco™ Expi293™ Expression Medium Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed as a core component to support high density culture of Expi293F™ cells in the Expi293™ Expression System for scalable transient protein expression
Gibco™ Collagenase, Type IV, Powder Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Cleaves bond between neutral amino acid (X) and glycine in sequence Pro-X-Glyc-Pro, which is found with high frequency in collagen
Gibco™ RPMI 1640 Medium, GlutaMAX™ Liquid Supplement, HEPES Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM/F-12, Powder, HEPES Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, Powder, HEPES is used in cell culture processing applications.
Gibco™ DMEM, High Glucose, GlutaMAX™ Supplement Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, GlutaMAX™ Supplement is used in human ex vivo tissue and cell culture processing applications.
Gibco™ Opti-MEM™ I Reduced Serum Medium, No Phenol Red Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Improved Minimal Essential Medium that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in cell growth rate or morphology
Gibco™ DMEM, Powder, Low Glucose, Pyruvate, No Sodium Bicarbonate Buffer Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, Powder, Low Glucose, Pyruvate, No Sodium Bicarbonate Buffer is used in cell culture processing applications.
Gibco™ MEM, No Glutamine, No Phenol Red, Liquid Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Minimum Essential Medium is commonly used of all cell culture media
Corning™ Lymphocyte Separation Medium Available on GSA/VA Contract for Federal Government customers only.
Designed for the in vitro isolation of lymphocytes from diluted whole blood.
Gibco™ RPMI 1640 Medium, No Phenol Red Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ DMEM, High Glucose, No Glutamine, No Phenol Red Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, No Glutamine, No Phenol Red is used in cell culture processing applications.
Gibco™ DMEM, High Glucose, HEPES Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, HEPES is used in cell culture processing applications.
Gibco™ IMDM, HEPES Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Iscove's Modified Dulbecco's Medium is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells
Gibco™ RPMI 1640 Medium, HEPES Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Originally developed to culture human leukemic cells in suspension and as monolayer
Corning™ Eagle's Minimum Essential Medium (MEM) Available on GSA/VA Contract for Federal Government customers only.
Used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts when supplemented with serum
  spinner