Vector Production Cell Culture Media
Invitrogen™ CSM Media for Mav203 Yeast Cells Non-distribution product as customer accommodation.
A specifically designed media for making agar plates that enable the growth of Saccharomyces cerevisae MaV203 cells
HyClone™ SFM4Transfx-293 Media
Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.
HyClone™ SFM4HEK293™ Media
Versatile, protein-free cell culture media, designed to support production of adenoviral vectors and growth of HEK 293 cultures.
EMD Millipore™ Novagen™ pTriEx™-1.1 Hygro DNA Available on GSA/VA Contract for Federal Government customers only.
Designed to allow rapid characterization of target genes in multiple expression systems. EMD Millipore™ Novagen™ pTriEx™-1.1 Hygro DNA can be used to test expression in E. coli, insect and vertebrate cells.
EMD Millipore™ Novagen™ pTriEx™-3 Neo DNA
Designed to allow rapid characterization of target genes in E. coli, insect and vertebrate cells. EMD Millipore™ Novagen™ pTriEx™-3 Neo DNA allows rapid selection of stable transfected vertebrate cells expressing high levels of target gene.
EMD Millipore™ Novagen™ pTriEx™-4 Neo DNA
Designed to allow rapid characterization of target genes in E. coli, insect and vertebrate cells. EMD Millipore™ Novagen™ pTriEx™-4 Neo DNA allows rapid selection of stable transfected vertebrate cells expressing high levels of target gene.
EMD Millipore™ Novagen™ pTriEx™-1.1 DNA
Designed to allow rapid characterization of target genes in multiple expression systems. EMD Millipore™ Novagen™ pTriEx™-1.1 DNA can be used to test expression in E. coli, insect and vertebrate cells.
EMD Millipore™ Novagen™ pTriEx™-6 DNA
Designed to allow rapid characterization of target genes in multiple expression systems. EMD Millipore™ Novagen™ pTriEx™-6 DNA can be used to test expression in E. coli, insect and mammalian cells.
EMD Millipore™ Novagen™ pTandem™-1 Vector
Coexpression of two genes from one vector in mammalian cells from a bicistronic RNA. EMD Millipore™ Novagen™ pTandem™-1 Vector contains separate multiple cloning sites located on both sides of an internal ribosome binding site (IRES).
EMD Millipore™ Novagen™ pTriEx™-5 DNA
Designed to allow rapid characterization of target genes in multiple expression systems. EMD Millipore™ Novagen™ pTriEx™-5 DNA can be used to test expression in E. coli, insect and mammalian cells.
EMD Millipore™ Novagen™ pTK-neo DNA
Used for selection of stably transfected mammalian cell lines (G418). EMD Millipore™ Novagen™ pTK-neo DNA has a minimal thymidine kinase (TK) promoter controls expression of the neomycin resistance gene.
EMD Millipore™ Novagen™ pTriEx™-4 DNA Available on GSA/VA Contract for Federal Government customers only.
Designed to allow rapid characterization of target genes in multiple expression systems. EMD Millipore™ Novagen™ pTriEx™-4 DNA can be used to test expression in E. coli, insect and vertebrate cells.
EMD Millipore™ Novagen™ pTriEx™-3 DNA
Designed to allow rapid characterization of target genes in multiple expression systems. EMD Millipore™ Novagen™ pTriEx™-3 DNA can be used to test expression in E. coli, insect and vertebrate cells.
HyClone™ SFM4Transfx 293 Media: PROMO
Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.
  spinner