Cell Culture Dishes
Thermo Scientific™ Nunc™ Non-Treated Multidishes Available on GSA/VA Contract for Federal Government customers only.
Perform suspension cell cultures with Thermo Scientific™ Nunc™ Non-Treated Multidishes.
Thermo Scientific™ Nunc™ Nunclon™ Vita 6-Well Multidish
Grow temperamental cells directly on the Thermo Scientific™ Nunc™ Nunclon™ Vita 6-Well Multidish without matrix coatings or feeder layers.
Thermo Scientific™ Nunc™ Cell Culture/Petri Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform a variety of applications across a broad range of sizes and styles to suit a variety of experimental needs.
Thermo Scientific™ Nunc™ MicroWell™ MiniTrays Available on GSA/VA Contract for Federal Government customers only.
Create more space in lab freezers and inubators with Nunc MicroWell MiniTrays, the optical clarity of which allows the trays to be used with inverted microscopes.
Thermo Scientific™ BioLite Cell Culture Treated Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform general cell culture procedures more economically with these dishes, featuring non-pyrogenic and non-cytotoxic cell-adherent surfaces.
Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface Available on GSA/VA Contract for Federal Government customers only.
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface.
Thermo Scientific™ Nunc™ Rectangular Dishes Available on GSA/VA Contract for Federal Government customers only.
Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.
Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes Available on GSA/VA Contract for Federal Government customers only.
Enhance cell attachment to the culture surface with the extracellular matrix coating on Thermo Scientific™ Nunc™ Poly-D-Lysine or Collagen I Coated Multidishes.
Thermo Scientific™ Nunc™ OmniTray™ Available on GSA/VA Contract for Federal Government customers only.
Screen libraries, DNA hybridization, standard petri dish techniques and bacterial growth with Thermo Scientific™ Nunc™ OmniTrays™.
Thermo Scientific™ Nunc™ Glass Bottom Dishes
Using Nunc Glass Bottom Dishes in your research applications gives you the flexibility to convert your culture device into an imaging device.
Thermo Scientific™ Nunclon™ Sphera™ Dishes
Features a superior cell culture surface with demonstrated performance that elevates stem cell and cell culture research.
Thermo Scientific™ Nunc™ MiniTrays with Nunclon™ Delta surface Available on GSA/VA Contract for Federal Government customers only.
Acheieve excellent cell attachment and growth rates with Nunc MiniTrays with Nunclon Delta Surface.
Corning™ BioCoat™ Fibronectin TC-treated Culture Dish
Matrix components are applied as an even, optically clear coating
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ Untreated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Binds bio-molecules through passive interactions and hydrophobic in nature. Corning™ Untreated Culture Dishes provide an ideal environment for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
Corning™ BioCoat™ Poly-D-Lysine/Laminin Culture Dish
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Corning™ BioCoat™ Matrigel™ Matrix Thin Layer Culture Dishes
Tested for the ability to promote neurite outgrowth from chick dorsal root ganglia (in the absence of NGF)
Corning™ BioCoat™ Laminin TC-treated Culture Dishes
Tested for ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ 60mm Center Well Culture Dish Available on GSA/VA Contract for Federal Government customers only.
Advanced center-well design promotes isolation, easy handling, and full visibility of cell
Corning™ BioCoat™ Gelatin Culture Dish
Pre-coating with gelatin (obtained from porcine gelatin) eliminates time-consuming preparation
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Corning™ CellBIND™ Surface Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Manufactured from optically-clear virgin polystyrene for excellent visibility. Corning™ CellBIND™ Suface Culture Dishes are treated for optimal cell attachment to help cells adapt to reduced serum or serum-free media.
Corning™ BioCoat™ Poly-L-Lysine Culture Dishes
Provide excellent substrate for cell attachment and differentiation of primary neurons, glial, neuroblastoma and glioma cells
Wheaton™ Culture Vessel
Perform repeated wet or dry sterilization to these culture vessels, which are made of borosilicate glass
Corning™ Ultra-Low Attachment Dishes Available on GSA/VA Contract for Federal Government customers only.
Unique covalently bonded hydrogel surface is hydrophilic and neutrally charged
Corning™ PureCoat™ Amine Culture Dishes
Positively charged surface improves attachment. increases proliferation and improved differentiation for a broad range of cells that exhibit poor attachment properties in serum-free or serum-reduced conditions
Corning™ BioCoat™ Poly-D-Lysine Culture Dishwith Coverslip Bottom
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Corning™ TC-Treated Culture Dishes Available on GSA/VA Contract for Federal Government customers only.
Tissue-culture (TC) treated for optimum attachment and growth of anchorage-dependent cells. Corning™ TC-Treated Culture Dishes provide an ideal solution for growing and maintaining cells in vitro.
  spinner