Cryogenic Storage Containers
Thermo Scientific™ Storage Boxes
Store a variety of vials and tubes with Thermo Scientific™ Storage Boxes, for use in ultra-low temperature freezers.
Thermo Scientific™ Cryoboxes Available on GSA/VA Contract for Federal Government customers only.
Meet any need and budget with Thermo Scientific™ Cryoboxes.
Corning™ CoolCell™ FTS30 Cell Freezing Vial Containers Available on GSA/VA Contract for Federal Government customers only.
Vials achieve a uniform and reproducible -1°C/minute freezing profile.
Corning™ CoolCell™ 5mL LX Cell Freezing Vial Container Available on GSA/VA Contract for Federal Government customers only.
Provides alcohol-free freezing at the rate of −1°C/minute when placed in a −80°C freezer.
Corning™ CoolCell™ LX Cell Freezing Vial Containers
Provides alcohol-free freezing at the rate of −1°C/minute when placed in a −80°C freezer.
Corning™ Cryogenic Vial Storage Boxes Available on GSA/VA Contract for Federal Government customers only.
For storage of internally and externally threaded vials in temperatures to –196°C (–320.8°F)
Thermo Scientific™ Storage Tube Protection Available on GSA/VA Contract for Federal Government customers only.
Protect tubes submerged in LN2 and prevent tube explosion with Thermo Scientific™ Storage Tube Protection.
Corning™ Cryopreservation Bags
Features a proprietary polyolefin/EVA (Ethyl Vinyl Acetate) film blend that remains flexible at ultra-low temperatures (-196°C)
Fisherbrand™ Cryogenic Vial Storage Boxes Available on GSA/VA Contract for Federal Government customers only.
Suitable for ultra-low temperature storage of both internally and externally threaded cryogenic vials
Argos Technologies PolarSafe™ Cryo/Freezer Standard Cardboard Box With Dividers
Used in mechanical freezers and liquid nitrogen tanks. Argos Technologies PolarSafe™ Cryo/Freezer Standard Cardboard Box With Dividers has small holes on bottom which allows safe drainage of liquid nitrogen or water.
Thermo Scientific™ Cryo Tube Holders Available on GSA/VA Contract for Federal Government customers only.
Simplify processes with these benchtop tube holders for single-handed capping and decapping.
Thermo Scientific™ Fiberboard Storage Boxes (Colored) Available on GSA/VA Contract for Federal Government customers only.
Store a variety of vials and tubes with Thermo Scientific™, water-repellant, fiberboard storage boxes, for use in ultra-low temperature or cryogenic freezers.
Corning™ CoolCell™ Cell Freezing Vial Containers
Controls rate of freezing to −1°C/minute when placed in a −80°C freezer.
Thermo Scientific™ Cryo Vial Closure Color Coders Available on GSA/VA Contract for Federal Government customers only.
Streamline the process of sample identification with Thermo Scientific™ Cryo Vial Closure Color Coders.
Argos Technologies™ PolarSafe™ Cryo/Freezer Mini Cardboard Box With 25-Place Cell Divider
Used in mechanical freezers and liquid nitrogen tanks. Argos Technologies PolarSafe™ Cryo/Freezer Mini Cardboard Box With 25-Place Cell Divider has small holes on bottom which allows safe drainage of liquid nitrogen or water.
Argos Technologies™ PolarSafe™ Cryo/Freezer Cardboard Box Without Cell Dividers
Used in mechanical freezers and liquid nitrogen tanks. Argos Technologies PolarSafe™ Cryo/Freezer Cardboard Box Without Cell Divider allows safe drainage of liquid nitrogen or water.
Thermo Scientific™ Nunc™ Storage Box and Rack Available on GSA/VA Contract for Federal Government customers only.
Hold up to 100 vials from 0.5 to 2.0mL in size in the Thermo Scientific™ Nunc™ Storage Box, or 24 vials in a 4x6 array in the Nunc Storage Racks.
Thermo Scientific™ Nunc™ CryoTube™ Colored Caps
Color-code your sample vials with colored replacement closures for Nunc internally-threaded CryoTube vials, featuring silicone gasket to provide the best seal.
Argos Technologies™ PolarSafe™ PCR Cardboard Freezer Boxes
Designed to hold single PCR tubes or strips of 8.
Thermo Scientific™ Storage Canes Available on GSA/VA Contract for Federal Government customers only.
Store various cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
Argos Technologies™ PolarSafe™ 81-Place Polypropylene Storage Boxes
Used for freezer storage and organization. Argos Technologies PolarSafe™ 81-Place Polypropylene Storage Boxes holds 0.5, 1.5, and 2.0mL microtubes as well as most brands of vials.
Fisherbrand™ Hinged Cryo/Freezer boxes - PROMO
Hinged Cryo/Freezer Boxes are sturdy and feature a hinged lid for easy sample access, eliminating swapped or misplaced box tops.
Fisherbrand™ Controlled Freeze Cubes - PROMO
Fisherbrand ontrolled Freeze Cubes have an open central chamber and aluminum alloy ring insert to aid the freezing process
Argos Technologies™ PolarSafe™ Colored Cardboard Freezer Boxes
For use with 81 and 100 place dividers. Argos Technologies PolarSafe™ Colored Cardboard Freezer Boxs fits most standard freezer racks.
Corning™ 2D Bar-coded Cryogenic Vial Storage Box for 81 Vials
For storing cryogenic vials. Corning™ 2D Bar-coded Cryogenic Vial Storage Box is made of polycarbonate.
DWK Life Sciences Wheaton™ KeepIT™ Freezer Boxes
Provides an ideal method for batching and storing samples.
Argos Technologies™ PolarSafe™ Cryo/Freezer Cell Dividers Only For Standard Tubes
Designed only for standard tubes. Argos Technologies PolarSafe™ Cryo/Freezer Cell Dividers Only For Standard Tubes has small holes on bottom which allows safe drainage of liquid nitrogen or water.
Bel-Art™ Cryo-Safe™ Vial Storage Boxes
Durable polycarbonate plastic boxes are excellent for compact storage of 1.2, 2.0 or 5.0mL cryogenic vials
Argos Technologies Magne-Box™ Polycarbonate Cryoboxes
For safe storage of cryovials. The Magne-Box™ Polycarbonate Cryobox has a magnetic lid closure which allows for one-handed retrievel from freezer racks.
Argos Technologies™ Gridless Polypropylene Storage Box
Designed to store larger tubes (lying flat) and laboratory vials. Argos Technologies Gridless Polypropylene Storage Box has venting holes in the bottom.
  spinner