Fetal Bovine Sera
Gibco™ Fetal Bovine Serum, Certified, Heat Inactivated, US Origin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Secure sera for cell culture with least viral risk sera from BSE-free regions
Corning™ Regular Fetal Bovine Serum Available on GSA/VA Contract for Federal Government customers only.
Supplement for cell culture
Gibco™ Fetal Bovine Serum, Qualified, USDA-approved Regions Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Trusted quality for consistent performance
Gibco™ Fetal Bovine Serum, Certified, US Origin, Standard (Sterile-Filtered) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Secure sera for cell culture with least viral risk sera from BSE-free regions
HyClone™ Fetal Bovine Serum (U.S.), Characterized
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
Gibco™ Fetal Bovine Serum, Qualified, Heat Inactivated, US Origin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Sera for specialty research and specific assays
Corning™ Premium Fetal Bovine Serum Available on GSA/VA Contract for Federal Government customers only.
Used as a supplement to cell culture media to help sustain growth and metabolism
Gibco™ Fetal Bovine Serum, Qualified, Heat Inactivated, USDA-approved Regions Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
General stem cell culture media and reagents; sera for specialty research and specific assays
HyClone™ Fetal Bovine Serum (U.S.), Defined
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
Gibco™ Fetal Bovine Serum, dialyzed , US Origin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Use for general cell culture with common cell lines
HyClone™ Fetal Bovine Serum (Canada), Characterized PROMO
Enhance cell culture growth with Canada sourced FBS tested for endotoxin, hemoglobin, and additional biochemical assays.
Gibco™ Fetal Bovine Serum, Qualified One Shot™, US origin Green Product Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Introducing the new, aliquot-free Gibco™ One Shot™ FBS 50 mL bottle. Designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it. And that improves your chances for consistent results.
Corning™ Fetal Bovine Serum, Premium (Gamma Irradiated) Available on GSA/VA Contract for Federal Government customers only.
Gamma irradiated FBS is sterile fetal bovine serum which is commonly used in vaccine and virus production.
GE Healthcare™ Characterized Fetal Bovine Serum, Canadian Origin PROMO
GE Healthcare™ Characterized Fetal Bovine Serum, Canadian Origin enhances the growth of cells with Canadian sourced HyClone Characterized Fetal Bovine Serum.
Gibco™ Fetal Bovine Serum, Qualified One Shot™, USDA Approved Regions Green Product Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Gibco™ sera: trusted quality for consistent performance
Gibco™ Fetal Bovine Serum, Certified One Shot™, US origin Green Product Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.
HyClone™ Fetal Bovine Serum (U.S.), Tetracyclin Screened
Maximize cell culture growth. HyClone™ Fetal Bovine Serum (U.S.), Tetracyclin Screened contains undetectable tetracycline levels and is designed for researchers using tetracycline-regulated gene expression in cultured cells, and filtered through sequential 40nm pore-size filters.
Gibco™ Heat Inactivated, Fetal Bovine Serum, Certified, ONE SHOT™, US Origin Non-distribution product as customer accommodation.
Heat-inactivated serum in a One-Shot 50-mL bottle increases workflow benefits, while maintaining serum quality. The 50-mL bottle eliminates the need to aliquot and reduces the risk of contamination.
Gibco™ One Shot™ Fetal Bovine Serum, Dialyzed, US Origin Non-distribution product as customer accommodation.
Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget require
Gibco™ Fetal Bovine Serum, ultra-low IgG, US Origin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
US-qualified performance sera for general cell culture with common cell lines
Gibco™ Fetal Bovine Serum, embryonic stem cell-Qualified, USDA-approved Regions Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
US certified performance plus sera for broad range of cell types, especially sensitive cell lines
Gibco™ One Shot™ Fetal Bovine Serum, Charcoal Strippped, US Origin Non-distribution product as customer accommodation.
Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget require
Corning™ Fetal Bovine Serum, Premium (Charcoal Stripped)
Charcoal stripped FBS has lower levels of lipophilic compounds such as hormones, growth factors, and steroids. This product is used by scientists conducting receptor or estrogen related studies which require lower levels of these components.
HyClone™ Fetal Bovine Serum (U.S.), Super Low IgG
Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production
  spinner