Other Sera
Alfa Aesar™ Platelet-poor Plasma Derived Serum, Bovine
For cellular and molecular biology applications
Gibco™ Bovine Serum, New Zealand Origin, Standard (Sterile-Filtered) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
US-qualified performance sera for general cell culture with common cell lines
Corning™ Donor Horse Serum, U.S. Sourced Available on GSA/VA Contract for Federal Government customers only.
Horse serum is often used in testing media for mycoplasma contamination or as a blocking agent and controls in immunoassays. It can also be combined with fetal bovine serum for the culture of rat adrenal pheochromocytoma (PC12) cells.
Gibco™ Bovine Serum, Heat Inactivated, New Zealand Origin Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Collected from cattle typically 12-24 months of age
HyClone™ FetalClone™ I Serum (U.S.)
Optimized for growth of hybridomas, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ I Serum, a fetal bovine serum (FBS) alternative containing comparable IgG levels to those found in FBS.
HyClone™ FetalClone™ II Serum (U.S.)
Developed for growth of CHO cells and derivatives, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ II Serum, a fetal bovine serum (FBS) alternative, containing additional growth factors and supplements than basic formulations.
HyClone™ Bovine Growth Serum (U.S.)
Support cell culture growth with U.S.-sourced HyClone™ Bovine Growth Serum, a superior performing fetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.
HyClone™ FetalClone™ III Serum (U.S.)
Designed for fibroblast applications, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ III Serum, the most widely applicable fetal bovine serum (FBS) alternative.
HyClone™ Donor Equine Serum (U.S.)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care.
SouthernBiotech™ Normal Donkey Serum
SouthernBiotech normal sera are useful as blocking reagents in a variety of immunoassays.
MilliporeSigma™ Calbiochem™ Normal Donkey Serum, Sterile Available on GSA/VA Contract for Federal Government customers only.
Suitable for use in radioimmunoassay. MilliporeSigma™; Calbiochem™ Normal Donkey Serum, Sterile is normal carrier sera, produced from pooled lots of sera from healthy, non-immunized donkeys.
MilliporeSigma™ Calbiochem™ Bovine γ-Globulin, Serum
Native γ-globulin from bovine serum.
Tocris Bioscience™ Bovine Serum Albumin Available on GSA/VA Contract for Federal Government customers only.
Widely used blocking agent for ELISA and blotting
HyClone™ FetalClone™ II Serum (U.S.) : PROMOTION
Promotion Details: Receive a 9&discount.
Supplement cell culture growth with a U.S. sourced FBS alternative, containing additional growth factors and supplements than basic formulations.
MP Biomedicals™ Whole Serum, Horse, Purified, 2mL
Whole Horse Serum with buffer salts
HyClone™ Donor Equine Serum
Carefully collected from selected U.S. donor herds that receive regular veterinary inspection and care.
GE Healthcare HyClone™ Cosmic Calf Serum (U.S. origin)
A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.
Novus Biologicals™ Donkey Whole Serum
Polyclonal serum without presevatives
GE Healthcare HyClone™ FetalClone™ II serum (U.S. origin)
Optimized for the growth of CHO cells and derivatives.
GE Healthcare HyClone™ Cosmic Calf Serum (New Zealand origin)
A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.
GE Healthcare HyClone™ Donor Equine Serum (U.S. origin)
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.
  spinner