Custom Enzymes and Inhibitors
BD General Caspase Inhibitor, Z-VAD-FMK Available on GSA/VA Contract for Federal Government customers only.
Cell permeable general caspase inhibitor that binds to the catalytic site of caspase proteases irreversibly and inhibits apoptosis
R&D Systems™ proTAME Available on GSA/VA Contract for Federal Government customers only.
Cell-permeable APC/C inhibitor
Tocris Bioscience™ ATPγS Tetralithium Salt Available on GSA/VA Contract for Federal Government customers only.
P2 agonist; ATP (Cat. No. 3245) analog
R&D Systems™ Epoxomicin
Cell-permeable, potent, selective and irreversible proteasome inhibitor
Thermo Scientific™ EpiJET™ Bisulfite Conversion Kit
The EpiJET Bisulfite Conversion Kit is used for an efficient bisulfite modification of DNA for methylation analysis.
BD Ac-DEVD-CHO Inhibitor Available on GSA/VA Contract for Federal Government customers only.
To identify and quantify Caspase-3 activity in apoptotic cells, and to study events downstream of Caspase-3 activation
BD Caspase-9 Inhibitor, Z-LEHD-FMK Available on GSA/VA Contract for Federal Government customers only.
Irreversible and cell permeable inhibitor of caspase-9
BD FUT-175 (Futhan) Available on GSA/VA Contract for Federal Government customers only.
Synthetic, broad-specificity protease inhibitor and an inhibitor of the classical and alternate pathways of complement activation
BD Caspase-8 Inhibitor, Z-IETD-FMKK Available on GSA/VA Contract for Federal Government customers only.
Irreversible and cell permeable inhibitor of caspase-8
BD Negative Control for Caspase Inhibitors, Z-FA-FMK Available on GSA/VA Contract for Federal Government customers only.
A negative control inhibitor which has no inhibitory effect on apoptosis mediated by caspases and can only inhibit cysteine proteases
Boston Biochem™ BostonBiochem UbiCREST Enzyme Set Available on GSA/VA Contract for Federal Government customers only.
Polyubiquitin chain linkage analysis system
BD Ac-IETD-CHO Caspase-8 Inhibitor Available on GSA/VA Contract for Federal Government customers only.
To study caspase-8 activity in apoptotic cell lysates
R&D Systems™ clasto-Lactacystin beta Lactone
Potent, highly specific, irreversible and cell-permeable inhibitor that covalently modifies the beta subunits of the proteasome
R&D Systems™ Z-Leu-Leu-Phe-CHO (Z-LLF-CHO)
Reversible, cell-permeable chymotrypsin-like peptidase inhibitor of the 20S proteasome
R&D Systems™ Me4BodipyFL-Ahx3Leu3VS (Proteasome Activity Probe)
A cell-permeable fluorescent proteasome activity probe for accurate profiling of proteasomal activity in cell lysates, intact cells, and murine and human patient-derived material, with high sensitivity
R&D Systems™ Z-Leu-Leu-Nva-CHO (MG-115)
Reversible, cell-permeable chymotrypsin-like peptidase inhibitor of the 20S proteasome
BD Caspase-3 Inhibitor, Z-DEVD-FMK Available on GSA/VA Contract for Federal Government customers only.
Irreversible and cell permeable inhibitor of caspase-3
R&D Systems™ b-AP15/VLX1500
Specifically inhibits the 19S regulatory particle-associated deubiquitinases USP14 and UCHL5, resulting in the accumulation of intracellular polyubiquitin
R&D Systems™ Z-Leu-Leu-Leu-B(OH)2 (MG-262) Available on GSA/VA Contract for Federal Government customers only.
Highly potent and selective cell-permeable inhibitor of the proteasome
R&D Systems™ Caspase Inhibitor Control Z-FA-FMK
Designed for in vitro use as a cell permeable control for the effects of fluoromethyl-ketone derivatives on caspase activity
R&D Systems™ PS-341, 26S Proteasome Inhibitor
A potent, specific, and reversible proteasome inhibitor
R&D Systems™ NEDD8 E1 Activating Enzyme Inhibitor Available on GSA/VA Contract for Federal Government customers only.
Contains the active component of MLN4924, a cell-permeable, small molecule inhibitor of Nedd8 activating enzyme
  spinner