Enzymes and Inhibitors
Thermo Scientific™ 50X TAE Buffer (Tris-acetate-EDTA) Available on GSA/VA Contract for Federal Government customers only.
Optimize electrophoresis of both genomic and large supercoiled DNA in agarose and polyacrylamide gels with Thermo Scientific™ 50X TAE Buffer (Tris-acetate-EDTA).
Thermo Scientific™ FastDigest™ NotI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest NotI restriction enzyme recognizes GC^GGCCGC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: CciNI).
Thermo Scientific™ FastDigest MssI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest™ Value Pack
FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in a single buffer.
Thermo Scientific™ FastDigest BsmBI (Esp3I) Available on GSA/VA Contract for Federal Government customers only.
Cut at CGTCTC(1/5)^ sites with Thermo Scientific™ FastDigest™ BsmBI (Esp3I), which peforms best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ Esp3I (BsmBI) Available on GSA/VA Contract for Federal Government customers only.
The Esp3I (BsmBI) restriction enzyme recognizes CGTCTC(1/5)^ sites and cuts best at 37°C in Tango (+DTT) buffer (Isoschizomers: BsmBI).
Thermo Scientific™ UTP Available on GSA/VA Contract for Federal Government customers only.
Optimize many molecular biology applications with UTP, a highly stable nucleotide with 99% purity.
Thermo Scientific™ FastDigest KpnI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ VspI (AseI) Available on GSA/VA Contract for Federal Government customers only.
Cut at AT^TAAT sites with VspI (AseI) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: AseI, PshBI).
Thermo Scientific™ FastDigest™ DpnI Available on GSA/VA Contract for Federal Government customers only.
Cut at Gm6A^TC sites with Thermo Scientific™ FastDigest™ DpnI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: MalI).
Thermo Scientific™ LguI (SapI) Available on GSA/VA Contract for Federal Government customers only.
The LguI (SapI) restriction enzyme recognizes GCTCTTC(1/4)^ sites and cuts best at 37°C in Tango buffer (Isoschizomers: BspQI, PciSI, SapI).
Thermo Scientific™ ATP Available on GSA/VA Contract for Federal Government customers only.
Optimize many molecular biology applications using this ATP, a highly stable nucleotide with 99% purity.
Thermo Scientific™ FastDigest NlaIII (Hin1II) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ GTP Available on GSA/VA Contract for Federal Government customers only.
Optimize molecular biology applications with this GTP, a highly stable nucleotide with 99% purity.
Thermo Scientific™ FastDigest Kpn2I Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ 10X TBE Buffer (Tris-borate-EDTA) Available on GSA/VA Contract for Federal Government customers only.
Perform electrophoresis of nucleic acids in agarose and polyacrylamide gels using Thermo Scientific™ 10X TBE Buffer (Tris-borate-EDTA).
Thermo Scientific™ FastDigest™ HindIII Available on GSA/VA Contract for Federal Government customers only.
The FastDigest HindIII restriction enzyme recognizes A^AGCTT sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ AarI Available on GSA/VA Contract for Federal Government customers only.
Cut at CACCTGC(4/8)^ sites with AarI restriction enzyme, which performs best at 37°C in its own unique (+oligo) buffer.
Thermo Scientific™ 10X Reaction Buffer with MgCl2 for DNase I Available on GSA/VA Contract for Federal Government customers only.
Optimize reaction condions for RNase-free DNase I with this 10X reaction Tris-HCl (pH 7.5) buffer containing MgCl2.
Thermo Scientific™ Van91I (PflMI) Available on GSA/VA Contract for Federal Government customers only.
Cut at CCANNNN^NTGG sites with Van91I (PflMI) restriction enzyme, which performs best at 37°C in R buffer (Isoschizomers: AccB7I, BasI, PflMI).
Thermo Scientific™ NTP Set Available on GSA/VA Contract for Federal Government customers only.
Optimize many molecular biology applications with this NTP set, composed of highly stable nucleotides with 99% purity.
Thermo Scientific™ I-SceI Available on GSA/VA Contract for Federal Government customers only.
Cut at TAGGGATAA^CAGGGTAAT sites with I-SceI homing enzyme, which performs best at 37°C in Tango buffer.
Thermo Scientific™ FastDigest NheI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ DpnI Available on GSA/VA Contract for Federal Government customers only.
Cut at Gm6A^TC sites with DpnI restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: MalI).
Thermo Scientific™ FastDigest™ NcoI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest NcoI restriction enzyme recognizes C^CATGG sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: Bsp19I).
Thermo Scientific™ KpnI Available on GSA/VA Contract for Federal Government customers only.
The KpnI restriction enzyme recognizes GGTAC^C sites and cuts best at 37°C in its own unique buffer (Isoschizomers: Asp718I).
Thermo Scientific™ FastDigest™ SpeI (BcuI) Available on GSA/VA Contract for Federal Government customers only.
FastDigest SpeI (BcuI) restriction enzyme recognizes A^CTAGT sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: AhlI).
Thermo Scientific™ FastDigest™ EcoRI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ FastDigest™ XhoI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Invitrogen™ DNase I, Amplification Grade Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Purified and tested for nondetectable levels of RNase contamination
  spinner