Aa - Bz Restriction Enzymes
Thermo Scientific™ FastDigest BsmBI (Esp3I) Available on GSA/VA Contract for Federal Government customers only.
Cut at CGTCTC(1/5)^ sites with Thermo Scientific™ FastDigest™ BsmBI (Esp3I), which peforms best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ AarI Available on GSA/VA Contract for Federal Government customers only.
Cut at CACCTGC(4/8)^ sites with AarI restriction enzyme, which performs best at 37°C in its own unique (+oligo) buffer.
Thermo Scientific™ ATP Available on GSA/VA Contract for Federal Government customers only.
Optimize many molecular biology applications using this ATP, a highly stable nucleotide with 99% purity.
Thermo Scientific™ FastDigest AgeI (BshTI) Available on GSA/VA Contract for Federal Government customers only.
Cut at A^CCGGT sites with Thermo Scientific™ FastDigest™ AgeI (BshTI), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ FastDigest BbsI (BpiI) Available on GSA/VA Contract for Federal Government customers only.
Cut at GAAGAC(2/6)^ sites with Thermo Scientific™ FastDigest™ BbsI (BpiI), which performs best using universal FastDigest Buffer (Isoschizomers: BpuAI, BstV2I).
Thermo Scientific™ BpiI (BbsI) Available on GSA/VA Contract for Federal Government customers only.
Cut GAAGAC(2/6)^ sites with BpiI (BbsI) restriction enzyme, which performs best at 37°C in G buffer (Isoschizomers: BbsI, BpuAI, BstV2I).
Thermo Scientific™ FastDigest™ BamHI Available on GSA/VA Contract for Federal Government customers only.
Cut at G^GATCC sites with Thermo Scientific™ FastDigest™ BamHI restriction enzyme, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ Bovine Serum Albumin (BSA) Available on GSA/VA Contract for Federal Government customers only.
Stabilize enzymes, increase PCR yields, and prevent adhesion of enzymes with Thermo Scientific™ Bovine Serum Albumin.
Thermo Scientific™ FastDigest™ BsmBI (Esp3I) Promotion Available on GSA/VA Contract for Federal Government customers only.
Cut at CGTCTC(1/5)^ sites with Thermo Scientific™ FastDigest™ BsmBI (Esp3I), which peforms best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Invitrogen™ Anza™ 4 BpiI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest AvrII (XmaJI) Available on GSA/VA Contract for Federal Government customers only.
Cut at C^CTAGG sites with Thermo Scientific™ FastDigest™ AvrII (XmaJI), which performs best using universal FastDigest Buffer (Isoschizomers: AspA2I, BlnI).
Thermo Scientific™ FastDigest AscI (SgsI) Available on GSA/VA Contract for Federal Government customers only.
Cut at GG^CGCGCC sites with Thermo Scientific™ FastDigest™ AscI (SgsI), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: PalAI).
Promega™ Restriction Enzymes - BstE II (D)
Star activity may be observed with glycerol concentrations >12% or with enzyme:DNA ratios >25u/μg.
Thermo Scientific™ AlfI
Cut at ^(10/12)GCA(N)6TGC(12/10)^ sites with AlfI restriction enzyme, which performs best at 37°C in R (+SAM) buffer.
Thermo Scientific™ FastDigest™ BbsI (BpiI) Promotion Available on GSA/VA Contract for Federal Government customers only.
Cut at GAAGAC(2/6)^ sites with Thermo Scientific™ FastDigest™ BbsI (BpiI), which performs best using universal FastDigest Buffer (Isoschizomers: BpuAI, BstV2I).
Thermo Scientific™ FastDigest BsiWI (Pfl23II) Available on GSA/VA Contract for Federal Government customers only.
Cut at C^GTACG sites with Thermo Scientific™ FastDigest™ BsiWI (Pfl23II), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: PspLI).
Thermo Scientific™ AdeI (DraIII) Available on GSA/VA Contract for Federal Government customers only.
Cut at CACNNN^GTG sites with AdeI (DraIII) restriction enzyme, which performs best at 37°C in G buffer (Isoschizomers: DraIII).
Thermo Scientific™ BamHI (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
Thermo Scientific™ FastDigest BstXI Available on GSA/VA Contract for Federal Government customers only.
Cut at CCANNNNN^NTGG sites with Thermo Scientific™ FastDigest™ BstXI, which performs best at 55°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ BcuI (SpeI) Available on GSA/VA Contract for Federal Government customers only.
Cut at A^CTAGT sites with BcuI (SpeI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: AhlI, SpeI).
Thermo Scientific™ BshTI (AgeI) Available on GSA/VA Contract for Federal Government customers only.
Cut at A^CCGGT sites with BshTI (AgeI) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: AgeI, AsiGI, CspAI, PinAI).
Thermo Scientific™ FastDigest ApaI Available on GSA/VA Contract for Federal Government customers only.
Cut at GGGCC^C sites with Thermo Scientific™ FastDigest™ ApaI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: PspOMI).
Promega™ Restriction Enzymes - BamH I Blue/White Cloning Qualified (E)
Capable of digesting DNA in 15 minutes or less. Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications.
Thermo Scientific™ FastDigest™ AvrII (XmaJI) Promotion
Cut at C^CTAGG sites with Thermo Scientific™ FastDigest™ AvrII (XmaJI), which performs best using universal FastDigest Buffer (Isoschizomers: AspA2I, BlnI).
Thermo Scientific™ FastDigest AatII Available on GSA/VA Contract for Federal Government customers only.
Cut at GACGT^C sites with Thermo Scientific™ FastDigest™ AatII, which performs best at 37°C in 5-15 minutes using universal FastDigest™ Buffer (Isoschizomers: ZraI).
Thermo Scientific™ FastDigest™ AatII Promotion
Cut at GACGT^C sites with Thermo Scientific™ FastDigest™ AatII, which performs best at 37°C in 5-15 minutes using universal FastDigest™ Buffer (Isoschizomers: ZraI).
Thermo Scientific™ BamHI Available on GSA/VA Contract for Federal Government customers only.
The BamHI restriction enzyme recognizes G^GATCC sites and cuts best at 37°C in its own unique buffer.
Thermo Scientific™ FastDigest BseGI Available on GSA/VA Contract for Federal Government customers only.
Cut at GGATG(2/0)^ sites with Thermo Scientific™ FastDigest™ BseGI performs best at 55°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: BstF5I, BtsCI, FokI).
Thermo Scientific™ BglII (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
  spinner