Ca -Dz Restriction Enzymes
Thermo Scientific™ Esp3I (BsmBI) Available on GSA/VA Contract for Federal Government customers only.
The Esp3I (BsmBI) restriction enzyme recognizes CGTCTC(1/5)^ sites and cuts best at 37°C in Tango (+DTT) buffer (Isoschizomers: BsmBI).
Thermo Scientific™ FastDigest™ DpnI Available on GSA/VA Contract for Federal Government customers only.
Cut at Gm6A^TC sites with Thermo Scientific™ FastDigest™ DpnI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: MalI).
Thermo Scientific™ FastDigest™ EcoRI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ DpnI Available on GSA/VA Contract for Federal Government customers only.
Cut at Gm6A^TC sites with DpnI restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: MalI).
Thermo Scientific™ Eco47III (AfeI) Available on GSA/VA Contract for Federal Government customers only.
The Eco47III (AfeI) restriction enzyme recognizes AGC^GCT sites and cuts best at 37°C in O buffer (Isoschizomers: AfeI, Aor51HI).
Invitrogen™ Anza™ 13 Esp3I Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ CpoI (RsrII) Available on GSA/VA Contract for Federal Government customers only.
The CpoI (RsrII) restriction enzyme recognizes CG^GWCCG sites and cuts best at 37°C in Tango buffer (Isoschizomers: CspI, Rsr2I, RsrII).
Thermo Scientific™ Cfr9I (XmaI) Available on GSA/VA Contract for Federal Government customers only.
The Cfr9I (XmaI) restriction enzyme recognizes C^CCGGG sites and cuts best at 37°C in its own unique buffer (Isoschizomers: TspMI, XmaCI, XmaI).
Thermo Scientific™ FastDigest™ Eco32I Available on GSA/VA Contract for Federal Government customers only.
Cut at GAT^ATC sites with Thermo Scientific™ FastDigest™ EcoRV (Eco32I), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: EcoRV).
Thermo Scientific™ EcoRI (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
Thermo Scientific™ FastDigest DdeI (HpyF3I) Available on GSA/VA Contract for Federal Government customers only.
Cut at C^TNAG sites with Thermo Scientific™ FastDigest™ DdeI (HpyF3I), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: BstDEI).
Invitrogen™ Anza™ 10 DpnI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ Eco32I (EcoRV) (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
Thermo Scientific™ Csp6I (CviQI) Available on GSA/VA Contract for Federal Government customers only.
Cut at G^TAC sites with Csp6I (CviQI) restriction enzyme, which performs best at 37°C in B buffer (Isoschizomers: AfaI, CviQI, RsaNI).
Thermo Scientific™ Eco31I (BsaI) Available on GSA/VA Contract for Federal Government customers only.
The Eco31I (BsaI) restriction enzyme recognizes GGTCTC(1/5)^ sites and cuts best at 37°C in G buffer (Isoschizomers: BsaI, Bso31I, BspTNI).
Thermo Scientific™ FastDigest FspAI Available on GSA/VA Contract for Federal Government customers only.
Cut at RTGC^GCAY sites with Thermo Scientific™ FastDigest™ FspAI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ Cfr42I (SacII) Available on GSA/VA Contract for Federal Government customers only.
The Cfr42I (SacII) restriction enzyme recognizes CCGC^GG sites and cuts best at 37°C in B buffer (Isoschizomers: KspI, SacII, Sfr303I, SgrBI, SstII).
Thermo Scientific™ EheI (SfoI) Available on GSA/VA Contract for Federal Government customers only.
Cut at GGC^GCC sites with EheI (SfoI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: BbeI, DinI, EgeI, KasI, Mly113I, NarI, SfoI).
Thermo Scientific™ CseI (HgaI) Available on GSA/VA Contract for Federal Government customers only.
Cut at GACGC(5/10)^ sites with CseI (HgaI) restriction enzyme, which performs best at 37°C in R buffer (Isoschizomers: HgaI).
Invitrogen™ Anza™ 11 EcoRI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ EcoRII Available on GSA/VA Contract for Federal Government customers only.
Cut at ^CCWGG sites with EcoRII restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: AjnI, BseBI, Bst2UI, BstNI, BstOI, Psp6I, PspGI).
Thermo Scientific™ Eco32I (EcoRV) Available on GSA/VA Contract for Federal Government customers only.
The Eco32I (EcoRV) restriction enzyme recognizes GAT^ATC sites and cuts best at 37°C in R buffer (Isoschizomers: EcoRV).
Thermo Scientific™ FastDigest™ ClaI (Bsu15I) Available on GSA/VA Contract for Federal Government customers only.
Cut at AT^CGAT sites with ClaI (Bsu15I) restriction enzyme, which performs best at 37°C in 5-15 minutes using universal FastDigest™ Buffer.
Thermo Scientific™ CTP Available on GSA/VA Contract for Federal Government customers only.
Optimize a variety of molecular biology applications with CTP, a highly stable nucleotide with 99% purity.
Thermo Scientific™ FastDigest DraI Available on GSA/VA Contract for Federal Government customers only.
Cut at TTT^AAA sites with Thermo Scientific™ FastDigest™ DraI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Promega™ Restriction Enzymes - EcoR I Blue/White Cloning Qualified (H)
Capable of digesting DNA in 15 minutes or less.
Thermo Scientific™ DraI Available on GSA/VA Contract for Federal Government customers only.
Cut at TTT^AAA sites with DraI restriction enzyme, which performs best at 37°C in Tango buffer.
Thermo Scientific™ FastDigest DraIII (AdeI) Available on GSA/VA Contract for Federal Government customers only.
Cut at CACNNN^GTG sites with Thermo Scientific™ FastDigest™ DraIII (AdeI), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ Eco147I (StuI) Available on GSA/VA Contract for Federal Government customers only.
The Eco147I (StuI) restriction enzyme recognizes AGG^CCT sites and cuts best at 37°C in B buffer (Isoschizomers: AatI, PceI, SseBI, StuI).
  spinner