Ea - Fz Restriction Enzymes
Thermo Scientific™ Esp3I (BsmBI) Available on GSA/VA Contract for Federal Government customers only.
The Esp3I (BsmBI) restriction enzyme recognizes CGTCTC(1/5)^ sites and cuts best at 37°C in Tango (+DTT) buffer (Isoschizomers: BsmBI).
Thermo Scientific™ FastDigest™ DpnI Available on GSA/VA Contract for Federal Government customers only.
Cut at Gm6A^TC sites with Thermo Scientific™ FastDigest™ DpnI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: MalI).
Thermo Scientific™ DpnI Available on GSA/VA Contract for Federal Government customers only.
Cut at Gm6A^TC sites with DpnI restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: MalI).
Thermo Scientific™ FastDigest™ EcoRI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ Cfr9I (XmaI) Available on GSA/VA Contract for Federal Government customers only.
The Cfr9I (XmaI) restriction enzyme recognizes C^CCGGG sites and cuts best at 37°C in its own unique buffer (Isoschizomers: TspMI, XmaCI, XmaI).
Thermo Scientific™ Eco31I (BsaI) Available on GSA/VA Contract for Federal Government customers only.
The Eco31I (BsaI) restriction enzyme recognizes GGTCTC(1/5)^ sites and cuts best at 37°C in G buffer (Isoschizomers: BsaI, Bso31I, BspTNI).
Thermo Scientific™ Csp6I (CviQI) Available on GSA/VA Contract for Federal Government customers only.
Cut at G^TAC sites with Csp6I (CviQI) restriction enzyme, which performs best at 37°C in B buffer (Isoschizomers: AfaI, CviQI, RsaNI).
Thermo Scientific™ FastDigest DraIII (AdeI) Available on GSA/VA Contract for Federal Government customers only.
Cut at CACNNN^GTG sites with Thermo Scientific™ FastDigest™ DraIII (AdeI), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ FastDigest™ ClaI (Bsu15I) Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Cut at AT^CGAT sites with ClaI (Bsu15I) restriction enzyme, which performs best at 37°C in 5-15 minutes using universal FastDigest™ Buffer.
Thermo Scientific™ FastDigest FspAI
Cut at RTGC^GCAY sites with Thermo Scientific™ FastDigest™ FspAI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ FastDigest EcoNI (XagI) Available on GSA/VA Contract for Federal Government customers only.
Cut at CCTNN^NNNAGG sites with Thermo Scientific™ FastDigest™ EcoNI (XagI), which performs best using universal FastDigest Buffer (Isoschizomers: BstENI).
Invitrogen™ Anza™ 13 Esp3I Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ CpoI (RsrII) Available on GSA/VA Contract for Federal Government customers only.
The CpoI (RsrII) restriction enzyme recognizes CG^GWCCG sites and cuts best at 37°C in Tango buffer (Isoschizomers: CspI, Rsr2I, RsrII).
Thermo Scientific™ DpnI (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
Thermo Scientific™ FastDigest™ Eco31I Available on GSA/VA Contract for Federal Government customers only.
The FastDigest Eco31I restriction enzyme recognizes GGTCTC(1/5)^ sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: BsaI, BspTNI).
Promega™ Restriction Enzymes - Dpn I (B)
Capable of digesting DNA in 15 minutes or less.
Thermo Scientific™ FastDigest DraI Available on GSA/VA Contract for Federal Government customers only.
Cut at TTT^AAA sites with Thermo Scientific™ FastDigest™ DraI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ FastDigest™ Eco32I Available on GSA/VA Contract for Federal Government customers only.
Cut at GAT^ATC sites with Thermo Scientific™ FastDigest™ EcoRV (Eco32I), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: EcoRV).
Thermo Scientific™ FastDigest EagI (Eco52I) Available on GSA/VA Contract for Federal Government customers only.
Cut at C^GGCCG sites with Thermo Scientific™ FastDigest™ EagI (Eco52I), which performs best universal FastDigest Buffer (Isoschizomers: BseX3I, BstZI, EclXI).
Thermo Scientific™ FastDigest DdeI (HpyF3I)
Cut at C^TNAG sites with Thermo Scientific™ FastDigest™ DdeI (HpyF3I), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: BstDEI).
Thermo Scientific™ FastDigest FspI (NsbI) Available on GSA/VA Contract for Federal Government customers only.
Cut at TGC^GCA sites with Thermo Scientific™ FastDigest™ FspI (NsbI), which performs best using universal FastDigest Buffer (Isoschizomers: Acc16I, AviII).
Thermo Scientific™ CTP Available on GSA/VA Contract for Federal Government customers only.
Optimize a variety of molecular biology applications with CTP, a highly stable nucleotide with 99% purity.
Thermo Scientific™ FastDigest Eam1105I Available on GSA/VA Contract for Federal Government customers only.
Cut at GACNNN^NNGTC sites with Thermo Scientific™ FastDigest™ Eam1105I, which performs best using universal FastDigest Buffer (Isoschizomers: AhdI, AspEI, BmeRI, DriI, EclHKI).
Thermo Scientific™ Cfr42I (SacII) (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
Thermo Scientific™ dm5CTP
Synthesize methylated DNA with the modified nucleotide dm5CTP.
Thermo Scientific™ Eco91I (BstEII) Available on GSA/VA Contract for Federal Government customers only.
The Eco91I (BstEII) restriction enzyme recognizes G^GTNACC sites and cuts best at 37°C in O buffer (Isoschizomers: BstEII, BstPI, EcoO65I, PspEI).
Thermo Scientific™ EcoRI Available on GSA/VA Contract for Federal Government customers only.
The EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37°C in its own unique buffer.
Thermo Scientific™ DraI Available on GSA/VA Contract for Federal Government customers only.
Cut at TTT^AAA sites with DraI restriction enzyme, which performs best at 37°C in Tango buffer.
Thermo Scientific™ CaiI (AlwNI)
Cut at CAGNNN^CTG sites with CaiI (AlwNI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: AlwNI).
  spinner