Ga - Hz Restriction Enzymes
Thermo Scientific™ GTP Available on GSA/VA Contract for Federal Government customers only.
Optimize molecular biology applications with this GTP, a highly stable nucleotide with 99% purity.
Thermo Scientific™ PasI Available on GSA/VA Contract for Federal Government customers only.
Cut at CC^CWGGG sites with PasI restriction enzyme, which performs best at 55°C in its own unique buffer.
Thermo Scientific™ FastDigest™ HindIII Available on GSA/VA Contract for Federal Government customers only.
The FastDigest HindIII restriction enzyme recognizes A^AGCTT sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ PfoI Available on GSA/VA Contract for Federal Government customers only.
Cut at T^CCNGGA sites with PfoI restriction enzyme recognizes and cuts best at 37°C in Tango buffer.
Thermo Scientific™ FastDigest HpaI (KspAI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest HincII Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest PacI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest HinfI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest RsaI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest™ PstI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest PstI restriction enzyme recognizes CTGCA^G sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: BspMAI).
Thermo Scientific™ PaeI (SphI) Available on GSA/VA Contract for Federal Government customers only.
The PaeI (SphI) restriction enzyme recognizes GCATG^C sites and cuts best at 37°C in B buffer (Isoschizomers: BbuI, SphI).
Thermo Scientific™ FastDigest HaeIII (BsuRI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ Hin1II (NlaIII) Available on GSA/VA Contract for Federal Government customers only.
Cut at CATG^ sites with Hin1II (NlaIII) restriction enzyme, which performs best at 37°C in G buffer (Isoschizomers: CviAII, FaeI, FatI, Hsp92II, NlaIII).
Thermo Scientific™ RsaI Available on GSA/VA Contract for Federal Government customers only.
The RsaI restriction enzyme recognizes GT^AC sites and cuts best at 37°C in Tango buffer (Isoschizomers: AfaI, CviQI, RsaNI).
Thermo Scientific™ PacI Available on GSA/VA Contract for Federal Government customers only.
The PacI restriction enzyme recognizes TTAAT^TAA sites and cuts best at 37°C in its own unique buffer.
Thermo Scientific™ FastDigest™ HindIII Promotion
The FastDigest HindIII restriction enzyme recognizes A^AGCTT sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ HindIII Available on GSA/VA Contract for Federal Government customers only.
The HindIII restriction enzyme recognizes A^AGCTT sites and cuts best at 37°C in R buffer.
Thermo Scientific™ PscI (PciI) Available on GSA/VA Contract for Federal Government customers only.
The PscI (PciI) restriction enzyme recognizes A^CATGT sites and cuts best at 37°C in Tango buffer (Isoschizomers: PciI).
Promega™ Restriction Enzymes - Hae III (C)
Capable of digesting DNA in 15 minutes or less.
Thermo Scientific™ FastDigest PvuII Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ HpyF3I (DdeI) Available on GSA/VA Contract for Federal Government customers only.
The HpyF3I (DdeI) restriction enzyme recognizes C^TNAG sites and cuts best at 37°C in Tango buffer (Isoschizomers: BstDEI, DdeI).
Thermo Scientific™ FastDigest PmlI (Eco72I) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Promega™ Restriction Enzymes - Hind III Blue/White Cloning Qualified (E)
Capable of digesting DNA in 15 minutes or less. Star activity may be observed in the presence of Mn2+.
Thermo Scientific™ FastDigest PsiI (AanI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest PvuI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Invitrogen™ Anza™ 16 HindIII Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest PshAI (BoxI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ GsuI (BpmI) Available on GSA/VA Contract for Federal Government customers only.
The GsuI (BpmI) restriction enzyme recognizes CTGGAG(16/14)^ sites and cuts best at 30°C in B buffer (Isoschizomers: BpmI).
Thermo Scientific™ FastDigest PdmI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest™ HpaI (KspAI) Promotion
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
  spinner