La - Mz Restriction Enzymes
Thermo Scientific™ LguI (SapI) Available on GSA/VA Contract for Federal Government customers only.
The LguI (SapI) restriction enzyme recognizes GCTCTTC(1/4)^ sites and cuts best at 37°C in Tango buffer (Isoschizomers: BspQI, PciSI, SapI).
Thermo Scientific™ FastDigest MssI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest MluI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ MboI Available on GSA/VA Contract for Federal Government customers only.
The MboI restriction enzyme recognizes ^GATC sites and cuts best at 37°C in R buffer (Isoschizomers: BfuCI, BssMI, BstKTI, BstMBI, DpnII, Kzo9I, NdeII, Sau3AIm).
Thermo Scientific™ MssI (PmeI) Available on GSA/VA Contract for Federal Government customers only.
The MssI (PmeI) restriction enzyme recognizes GTTT^AAAC sites and cuts best at 37°C in B buffer (Isoschizomers: PmeI).
Thermo Scientific™ MreI (Sse232I) Available on GSA/VA Contract for Federal Government customers only.
Cut at CG^CCGGCG sites with MreI (Sse232I) restriction enzyme, which performs best at 37°C in G buffer.
Invitrogen™ Anza™ 10 DpnI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Promega™ Restriction Enzymes - Mbo I (C)
Sensitive to dam methylation. Sau3Al and Dpnl are isoschizomers of Mbol.
Thermo Scientific™ MspI (HpaII) Available on GSA/VA Contract for Federal Government customers only.
The MspI (HpaII) restriction enzyme recognizes C^CGG sites and cuts best at 37°C in Tango buffer.
Thermo Scientific™ FastDigest™ MunI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Invitrogen™ Anza™ 31 MunI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest MscI (MlsI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest MseI (SaqAI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Invitrogen™ Anza™ 24 MssI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 33 LguI Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest MreI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest MauBI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ MluI Available on GSA/VA Contract for Federal Government customers only.
The MluI restriction enzyme recognizes A^CGCGT sites and cuts best at 37°C in R buffer.
Thermo Scientific™ MauBI Available on GSA/VA Contract for Federal Government customers only.
Cut T CG^CGCGCG sites with MauBI restriction enzyme, which performs best at 37°C in Tango buffer.
Thermo Scientific™ FastDigest Mva1269I Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Promega™ Restriction Enzymes - Msp I (B)
MspI and its isoschizomer HpaII exhibit different methylation sensitivities.
Thermo Scientific™ MvaI (BstNI) Available on GSA/VA Contract for Federal Government customers only.
Cut at CC^WGG sites with MvaI (BstNI) restriction enzyme, which performs best at 37°C in R buffer (Isoschizomers: AjnI, BseBI, Bst2UI, BstNI, BstOI, Psp6I, PspGI).
Thermo Scientific™ FastDigest MlyI (SchI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Invitrogen™ Anza™ 37 Mph1103I Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ Mph1103I (NsiI) Available on GSA/VA Contract for Federal Government customers only.
The Mph1103I (NsiI) restriction enzyme recognizes ATGCA^T sites and cuts best at 37°C in R buffer (Isoschizomers: EcoT22I, NsiI, Zsp2I).
Thermo Scientific™ MboII
The MboII restriction enzyme recognizes GAAGA(8/7)^ sites and cuts best at 37°C in B buffer.
Invitrogen™ Anza™ 28 MluI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 85 MreI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ MunI (MfeI) Available on GSA/VA Contract for Federal Government customers only.
The MunI (MfeI) restriction enzyme recognizes C^AATTG sites and cuts best at 37°C in G buffer (Isoschizomers: MfeI).
Thermo Scientific™ MlsI (MscI) Available on GSA/VA Contract for Federal Government customers only.
The MlsI (MscI) restriction enzyme recognizes TGG^CCA sites and cuts best at 37°C in R buffer (Isoschizomers: BalI, MluNI, MscI, Msp20I).
  spinner