Pa - Rz Restriction Enzymes
Thermo Scientific™ FastDigest™ NotI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest NotI restriction enzyme recognizes GC^GGCCGC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: CciNI).
Thermo Scientific™ FastDigest NheI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ NTP Set Available on GSA/VA Contract for Federal Government customers only.
Optimize many molecular biology applications with this NTP set, composed of highly stable nucleotides with 99% purity.
Thermo Scientific™ PasI Available on GSA/VA Contract for Federal Government customers only.
Cut at CC^CWGGG sites with PasI restriction enzyme, which performs best at 55°C in its own unique buffer.
Thermo Scientific™ FastDigest™ NcoI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest NcoI restriction enzyme recognizes C^CATGG sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: Bsp19I).
Thermo Scientific™ FastDigest PacI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ PfoI Available on GSA/VA Contract for Federal Government customers only.
Cut at T^CCNGGA sites with PfoI restriction enzyme recognizes and cuts best at 37°C in Tango buffer.
Thermo Scientific™ FastDigest PvuI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest RsaI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ NotI Available on GSA/VA Contract for Federal Government customers only.
The NotI restriction enzyme recognizes GC^GGCCGC sites and cuts best at 37°C in O buffer (Isoschizomers: CciNI).
Invitrogen™ Anza™ 1 NotI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest™ NdeI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest NdeI restriction enzyme recognizes CA^TATG sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: FauNDI).
Thermo Scientific™ PaeI (SphI) Available on GSA/VA Contract for Federal Government customers only.
The PaeI (SphI) restriction enzyme recognizes GCATG^C sites and cuts best at 37°C in B buffer (Isoschizomers: BbuI, SphI).
Thermo Scientific™ FastDigest™ PstI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest PstI restriction enzyme recognizes CTGCA^G sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: BspMAI).
Thermo Scientific™ RsaI Available on GSA/VA Contract for Federal Government customers only.
The RsaI restriction enzyme recognizes GT^AC sites and cuts best at 37°C in Tango buffer (Isoschizomers: AfaI, CviQI, RsaNI).
Thermo Scientific™ FastDigest NheI
Thermo Scientific FastDigest NheI restriction enzyme recognizes G^CTAGC site and cuts best at 37°C in 5–15 minutes using universal FastDigest Buffer.
Thermo Scientific™ FastDigest PsiI (AanI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest NsiI (Mph1103I) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest PvuII Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ PacI Available on GSA/VA Contract for Federal Government customers only.
The PacI restriction enzyme recognizes TTAAT^TAA sites and cuts best at 37°C in its own unique buffer.
Thermo Scientific™ FastDigest PmlI (Eco72I) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ PscI (PciI) Available on GSA/VA Contract for Federal Government customers only.
The PscI (PciI) restriction enzyme recognizes A^CATGT sites and cuts best at 37°C in Tango buffer (Isoschizomers: PciI).
Thermo Scientific™ PfeI (TfiI) Available on GSA/VA Contract for Federal Government customers only.
Cut at G^AWTC sites with PfeI (TfiI) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: TfiI).
Thermo Scientific™ NheI Available on GSA/VA Contract for Federal Government customers only.
The NheI restriction enzyme recognizes G^CTAGC sites and cuts best at 37°C in Tango buffer (Isoschizomers: AsuNHI, BmtI).
Promega™ Restriction Enzymes - Nde I (D)
Active and capable of digesting DNA in 15 minutes or less, directly in GoTaq™ Green Master Mix.
Thermo Scientific™ FastDigest PdmI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ NdeI (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
Thermo Scientific™ FastDigest™ NotI Promotion
The FastDigest NotI restriction enzyme recognizes GC^GGCCGC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: CciNI).
  spinner