Sa - Tz Restriction Enzymes
Thermo Scientific™ 50X TAE Buffer (Tris-acetate-EDTA) Available on GSA/VA Contract for Federal Government customers only.
Optimize electrophoresis of both genomic and large supercoiled DNA in agarose and polyacrylamide gels with Thermo Scientific™ 50X TAE Buffer (Tris-acetate-EDTA).
Thermo Scientific™ 10X TBE Buffer (Tris-borate-EDTA) Available on GSA/VA Contract for Federal Government customers only.
Perform electrophoresis of nucleic acids in agarose and polyacrylamide gels using Thermo Scientific™ 10X TBE Buffer (Tris-borate-EDTA).
Invitrogen™ Anza™ 14 SalI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest SapI (LguI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest SalI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ SgrDI Available on GSA/VA Contract for Federal Government customers only.
Cut at CG^TCGACG sites with SgrDI restriction enzyme, which functions best at 37°C in R buffer.
Thermo Scientific™ FastDigest SwaI (SmiI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest SanDI (KflI)
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ SmiI (SwaI) Available on GSA/VA Contract for Federal Government customers only.
Cut at ATTT^AAAT sites with SmiI (SwaI) restriction enzyme, which performs best at 30°C in O buffer (Isoschizomers: SwaI).
Thermo Scientific™ FastDigest SbfI (SdaI) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ 10X T4 DNA Ligase Buffer Available on GSA/VA Contract for Federal Government customers only.
Safely ligate samples with this buffer, for use with T4 DNA Ligase and tested for the absence of endo- and exodeoxyribonucleases and ribonucleases.
Thermo Scientific™ TaqI (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
Thermo Scientific™ FastDigest™ SpeI (BcuI) Available on GSA/VA Contract for Federal Government customers only.
FastDigest SpeI (BcuI) restriction enzyme recognizes A^CTAGT sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: AhlI).
Promega™ Restriction Enzymes - Sgf I Genome Qualified (C)
Genome qualified to ensure optimal performance in genomic analysis applications. Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications.
Promega™ Restriction Enzymes - Sca I (K)
Capable of digesting DNA in 15 minutes or less. Available at high concentration, containing 5,000 units of ScaI at a concentration of 40-80u/μL
Thermo Scientific™ FastDigest™ SacI Available on GSA/VA Contract for Federal Government customers only.
The FastDigest SacI restriction enzyme recognizes GAGCT^C sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: Eco53kI, EcoICRI, Psp124BI, SstI)
Thermo Scientific™ FastDigest StuI (Eco147I) Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ SspDI (KasI) Available on GSA/VA Contract for Federal Government customers only.
Cut at G^GCGCC sites with SspDI (KasI) restriction enzyme, which functions best at 37°C in Tango buffer (Isoschizomers: BbeI, DinI, EgeI, KasI, Mly113I, NarI, SfoI).
Thermo Scientific™ FastDigest SspI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ 10X Taq™ Available on GSA/VA Contract for Federal Government customers only.
Find the right buffer to meet your specific PCR needs in these 10X Taq DNA Polymerase buffers with different compositions.
Promega SmaI, 5,000u
CCC/GGG GGG\CCC
Thermo Scientific™ SgsI (AscI) Available on GSA/VA Contract for Federal Government customers only.
The SgsI (AscI) restriction enzyme recognizes GG^CGCGCC sites and cuts best at 37°C in Tango buffer (Isoschizomers: AscI, PalAI).
Thermo Scientific™ SalI Available on GSA/VA Contract for Federal Government customers only.
The SalI restriction enzyme recognizes G^TCGAC sites and cuts best at 37°C in O buffer.
Thermo Scientific™ FastDigest ScaI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Promega™ Restriction Enzymes - Sma I Blue/White Cloning and Genome Qualified (J)
XmaI is a neoschizomer of SmaI. Available at high concentration, containing 5,000 units of SmaI at a concentration of 40-80u/μL.
Thermo Scientific™ TatI Available on GSA/VA Contract for Federal Government customers only.
Cut at W^GTACW sites with TatI restriction enzyme, which performs best at 65°C in Tango buffer.
Thermo Scientific™ FastDigest TauI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ TasI (Tsp509I) Available on GSA/VA Contract for Federal Government customers only.
Cut at ^AATT sites with TasI (Tsp509I) restriction enzyme, which performs best at 65°C in B buffer (Isoschizomers: Sse9I, Tsp509I, TspEI).
Thermo Scientific™ SfiI Available on GSA/VA Contract for Federal Government customers only.
Cut at GGCCNNNN^NGGCC sites with SfiI restriction enzyme, which performs best at 50°C in G buffer.
Thermo Scientific™ TaaI (HpyCH4III) Available on GSA/VA Contract for Federal Government customers only.
The TaaI (HpyCH4III) restriction enzyme recognizes ACN^GT sites and cuts best at 65°C in Tango buffer (Isoschizomers: Bst4CI, HpyCH4III).
  spinner