Ua - Wz Restriction Enzymes
Thermo Scientific™ FastDigest™ Value Pack
FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in a single buffer.
Thermo Scientific™ UTP Available on GSA/VA Contract for Federal Government customers only.
Optimize many molecular biology applications with UTP, a highly stable nucleotide with 99% purity.
Thermo Scientific™ VspI (AseI) Available on GSA/VA Contract for Federal Government customers only.
Cut at AT^TAAT sites with VspI (AseI) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: AseI, PshBI).
Thermo Scientific™ Van91I (PflMI) Available on GSA/VA Contract for Federal Government customers only.
Cut at CCANNNN^NTGG sites with Van91I (PflMI) restriction enzyme, which performs best at 37°C in R buffer (Isoschizomers: AccB7I, BasI, PflMI).
Invitrogen™ Anza™ 106 Van91I Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 76 VspI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
  spinner