Xa - Zz Restriction Enzymes
Thermo Scientific™ FastDigest™ XhoI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest™ XbaI Available on GSA/VA Contract for Federal Government customers only.
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest™ XhoI Promotion
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ XmaJI (AvrII) Available on GSA/VA Contract for Federal Government customers only.
Cut at C^CTAGG sites with XmaJI (AvrII) restriction enzyme, which functions best at 37°C in Tango buffer (Isoschizomers: AspA2I, AvrII, BlnI).
Thermo Scientific™ XbaI (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System
Thermo Scientific™ FastDigest™ XbaI Promotion
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ XhoI Available on GSA/VA Contract for Federal Government customers only.
Cut at C^TCGAG sites with XhoI restriction enzyme, which functions best at 37°C in R buffer (Isoschizomers: PaeR7I, Sfr274I, SlaI, StrI, TliI).
Promega™ Restriction Enzymes - Xba I Blue/White Cloning and Genome Qualified (D)
Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications.
Thermo Scientific™ XhoI (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
Thermo Scientific™ XbaI Available on GSA/VA Contract for Federal Government customers only.
Cut at T^CTAGA sites with XbaI restriction enzyme, which functions best at 37°C in Tango buffer.
Invitrogen™ Anza™ 8 XhoI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ XapI (ApoI) Available on GSA/VA Contract for Federal Government customers only.
Cut at R^AATTY sites with XapI (ApoI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: AcsI, ApoI).
Thermo Scientific™ FastDigest XapI Available on GSA/VA Contract for Federal Government customers only.
FastDigest XapI restriction enzyme recognizes R^AATTY sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: AcsI, ApoI).
Invitrogen™ Anza™ 12 XbaI Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ XagI (EcoNI) Available on GSA/VA Contract for Federal Government customers only.
Cut at CCTNN^NNNAGG sites with XagI (EcoNI) restriction enzyme, which performs best at 37°C in R buffer (Isoschizomers: BstENI, EcoNI).
Thermo Scientific™ XmiI (AccI) Available on GSA/VA Contract for Federal Government customers only.
Cut at GT^MKAC sites with XmiI (AccI) restriction enzyme, which functions best at 37°C in B buffer (Isoschizomers: AccI, FblI).
Promega™ Restriction Enzymes - Xho I Blue/White Cloning and Genome Qualified (D)
Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications.
Thermo Scientific™ FastDigest™ XapI Promotion
FastDigest XapI restriction enzyme recognizes R^AATTY sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: AcsI, ApoI).
Thermo Scientific™ XceI (NspI) Available on GSA/VA Contract for Federal Government customers only.
Cut at RCATG^Y sites with XceI (NspI) restriction enzyme, which performs best at 37°C in Tango buffer (Isoschizomers: BstNSI, NspI).
FastDigest™ XapI Restriction Enzyme
Recognizes R^AATTY sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: AcsI, ApoI).
Thermo Scientific™ XceI (NspI) (10 U/µL)
Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System.
FastDigest™ XapI Promotion
FastDigest XapI restriction enzyme recognizes R^AATTY sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: AcsI, ApoI).
Invitrogen™ Anza™ 99 XagI Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 96 XmiI Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 98 XceI Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 111 XapI Non-distribution product as customer accommodation.
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
  spinner