Autoradiography Supplies
Thermo Scientific™ Pierce™ PVDF Membranes Available on GSA/VA Contract for Federal Government customers only.
Enable high protein and nucleic acid binding capacity with sheets and rolls of high-quality polyvinylidene difluoride (PVDF) membrane.
Thermo Scientific™ CL-XPosure™ Film (X-Ray Film) Available on GSA/VA Contract for Federal Government customers only.
Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.
Thermo Scientific™ Pierce™ Western Blotting Filter Papers Available on GSA/VA Contract for Federal Government customers only.
Pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels to PVDF, nitrocellulose, or other membranes.
GE Healthcare Amersham™ Hypercassette™ Autoradiography Cassette Available on GSA/VA Contract for Federal Government customers only.
Use for the toughest demands of research in the life sciences. GE Healthcare Life Sciences™ Amersham™ Hypercassette™ Autoradiography Cassettes are constructed from strong aluminium and a choice of colors enables color coding of work.
Thermo Scientific™ Pierce™ Nitrocellulose Membranes
Sheets and rolls of nitrocellulose, available in two pore sizes, with excellent binding properties for Western blot and other protein or nucleic acid methods.
GE Healthcare Amersham™ Hyperfilm™ ECL Available on GSA/VA Contract for Federal Government customers only.
Use with all blue and green light chemiluminescent systems. GE Healthcare Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
Carestream Health BioMax™ MS Autoradiography Intensifying Screens
Conventional format intensifying screens for maximum sensitivity in detection of higher energy radioisotopes such as 32P and 125I
Thermo Scientific™ Pierce™ 1-Step Transfer Buffer Available on GSA/VA Contract for Federal Government customers only.
Perform rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
GE Healthcare Amersham™ Hyperscreen™ Intensifying Screen Available on GSA/VA Contract for Federal Government customers only.
Offers rapid results and high sensitivity for detecting 32P and 125I in Southern, Northern, and Western blots
Carestream Health BioMax™ TranScreen™ LE Autoradiography Intensifying Screens
Intensifying screen designed for 3H, 14C, 33P, 35S, 45Ca, and 59Fe
Thermo Scientific™ Pierce™ Nylon Membranes Available on GSA/VA Contract for Federal Government customers only.
Sheets of unmodified or positively charged Biodyne A and Biodyne B Nylon that provide excellent binding characteristics for nucleic acid transfer, blotting and detection.
Bel-Art™ SP Scienceware™ X-Ray Cassette Security Bags
Intended to augment the light protection afforded by X-ray film cassettes which may leak light because of damage due to age, use, or sudden shock
Carestream Health BioMax™ MR (Maximum Resolution) Film
Recommended for direct autoradiography of weak and medium energy β-radioisotopes, such as 35S,33P, and 14C
GE Healthcare Amersham™ Hyperfilm™ MP Available on GSA/VA Contract for Federal Government customers only.
Use in automatic processors or manually in tanks and trays using most common X-ray film developers and fixers. GE Healthcare Life Sciences™ Amersham™ Hyperfilm™ MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti scratch layer.
Thermo Scientific™ Pierce™ Western Blot Roller
A simple, reusable tool for gently and easily removing trapped air bubbles when assembling transfer stacks in semi-dry or wet-tank transfer cassettes.
Carestream Health BioMax™ XAR™ Film
Suitable for use with all radioisotopes and chemiluminescent probes
Thermo Scientific™ Pierce™ Background Eliminator for Film
Rapidly remove haze and artifacts from X-ray film to correct overexposed blots, whether from chemiluminescent Western blot or DNA or RNA autoradiography.
Carestream Health BioMax™ Light Film
Developed specifically for the detection of chemiluminescent probes
Carestream Health BioMax™ MS (Maximum Sensitivity) Film
Ideal for use with radioisotopes when detection of low-intensity samples or reduction of exposure times is required
Thermo Scientific™ Pierce™ Antibody Extender Solution NC
Enable Western blot detection with much less primary antibody by treating nitrocellulose membrane with this reagent.
Carestream Health X-Omat™ LS Film
Economical, blue-tinted film is suitable for use with all radioisotopes and chemiluminescent probes
Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts
Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.
Thermo Scientific™ myImageAnalysis™ Software
myImageAnalysis Software is a full-featured, multifunction computer program to analyze and edit digital images of electrophoresis gels and blots that were acquired using gel documentation imagers or scanners.
Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter
These electrical power cords have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.
Minigrip™ Protex-Ray™ Clear Film Disposable X-Ray Cassette Covers
Helps reduce the risk of contamination when handling cassettes for portable Xx-rays
Carestream Health BioMax™ TranScreen™ HE Autoradiography Intensifying Screens
Innovative intensifying screen developed specifically for detection of higher energy radioisotopes such as 32P and 125I
  spinner