Autoradiography Cassettes and Screens
GE Healthcare Amersham™ Hypercassette™ Autoradiography Cassette Available on GSA/VA Contract for Federal Government customers only.
Use for the toughest demands of research in the life sciences. GE Healthcare Life Sciences™ Amersham™ Hypercassette™ Autoradiography Cassettes are constructed from strong aluminium and a choice of colors enables color coding of work.
GE Healthcare Amersham™ Hyperscreen™ Intensifying Screen Available on GSA/VA Contract for Federal Government customers only.
Offers rapid results and high sensitivity for detecting 32P and 125I in Southern, Northern, and Western blots
Bel-Art™ SP Scienceware™ X-Ray Cassette Security Bags Available on GSA/VA Contract for Federal Government customers only.
Intended to augment the light protection afforded by X-ray film cassettes which may leak light because of damage due to age, use, or sudden shock
Minigrip™ Protex-Ray™ Clear Film Disposable X-Ray Cassette Covers
Helps reduce the risk of contamination when handling cassettes for portable Xx-rays
Carestream Health BioMax™ MS Autoradiography Intensifying Screens
Conventional format intensifying screens for maximum sensitivity in detection of higher energy radioisotopes such as 32P and 125I
Carestream Health BioMax™ TranScreen™ LE Autoradiography Intensifying Screens
Intensifying screen designed for 3H, 14C, 33P, 35S, 45Ca, and 59Fe
Carestream Health BioMax™ TranScreen™ HE Autoradiography Intensifying Screens
Innovative intensifying screen developed specifically for detection of higher energy radioisotopes such as 32P and 125I
Fisherbrand™ Intensifying Screens
Decrease exposure times and lower levels of radioactivity needed for autoradiography of gels
Carestream Health BioMax™ Autoradiography Cassettes
Maximize image resolution and background clarity by ensuring intimate contact between the film and sample in a light-tight environment
  spinner