Autoradiography Film
Thermo Scientific™ CL-XPosure™ Film (X-Ray Film) Available on GSA/VA Contract for Federal Government customers only.
Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.
Thermo Scientific™ Pierce™ PVDF Membranes Available on GSA/VA Contract for Federal Government customers only.
Enable high protein and nucleic acid binding capacity with sheets and rolls of high-quality polyvinylidene difluoride (PVDF) membrane.
Thermo Scientific™ Pierce™ Western Blotting Filter Papers Available on GSA/VA Contract for Federal Government customers only.
Pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels to PVDF, nitrocellulose, or other membranes.
Thermo Scientific™ Pierce™ Nitrocellulose Membranes Available on GSA/VA Contract for Federal Government customers only.
Sheets and rolls of nitrocellulose, available in two pore sizes, with excellent binding properties for Western blot and other protein or nucleic acid methods.
GE Healthcare Amersham™ Hyperfilm™ ECL Available on GSA/VA Contract for Federal Government customers only.
Use with all blue and green light chemiluminescent systems. GE Healthcare Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
Thermo Scientific™ Pierce™ Nylon Membranes Available on GSA/VA Contract for Federal Government customers only.
Sheets of unmodified or positively charged Biodyne A and Biodyne B Nylon that provide excellent binding characteristics for nucleic acid transfer, blotting and detection.
GE Healthcare Amersham™ Hyperfilm™ MP Available on GSA/VA Contract for Federal Government customers only.
Use in automatic processors or manually in tanks and trays using most common X-ray film developers and fixers. GE Healthcare Life Sciences™ Amersham™ Hyperfilm™ MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti scratch layer.
Thermo Scientific™ Pierce™ 1-Step Transfer Buffer Available on GSA/VA Contract for Federal Government customers only.
Perform rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Pierce G2 Fast Blotter.
Carestream Health BioMax™ MR (Maximum Resolution) Film
Recommended for direct autoradiography of weak and medium energy β-radioisotopes, such as 35S,33P, and 14C
Carestream Health BioMax™ Light Film
Developed specifically for the detection of chemiluminescent probes
Thermo Scientific™ Pierce™ Western Blot Roller
A simple, reusable tool for gently and easily removing trapped air bubbles when assembling transfer stacks in semi-dry or wet-tank transfer cassettes.
Carestream Health BioMax™ XAR™ Film
Suitable for use with all radioisotopes and chemiluminescent probes
GE Healthcare Amersham™ Hyperfilm™ ECL: PROMO
Use with all blue and green light chemiluminescent systems. GE Healthcare Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
Carestream Health X-Omat™ LS Film
Economical, blue-tinted film is suitable for use with all radioisotopes and chemiluminescent probes
Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts
Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.
Thermo Scientific™ Pierce™ Background Eliminator for Film
Rapidly remove haze and artifacts from X-ray film to correct overexposed blots, whether from chemiluminescent Western blot or DNA or RNA autoradiography.
Carestream Health BioMax™ MS (Maximum Sensitivity) Film
Ideal for use with radioisotopes when detection of low-intensity samples or reduction of exposure times is required
Thermo Scientific™ Pierce™ Antibody Extender Solution NC
Enable Western blot detection with much less primary antibody by treating nitrocellulose membrane with this reagent.
GE Healthcare Amersham™ Hyperfilm™ MP: PROMO
Use in automatic processors or manually in tanks and trays using most common X-ray film developers and fixers. GE Healthcare Amersham™ Hyperfilm™ MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti scratch layer.
Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter
These electrical power cords have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.
Thermo Scientific™ myImageAnalysis™ Software
myImageAnalysis Software is a full-featured, multifunction computer program to analyze and edit digital images of electrophoresis gels and blots that were acquired using gel documentation imagers or scanners.
Thermo Scientific™ myECL™ Imager
One-touch image capture and analysis for protein and nucleic acid gels and blots
Thermo Scientific™ Pierce™ G2 Fast Blotter
Designed specifically for rapid semi-dry transfer of 10-300kDa proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 10 minutes.
GE Healthcare GE Healthcare Amersham™ Hyperfilm™ ECL Promo
Superior contrast and improved band visibility
  spinner