Flow Cytometry Supplies
BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution Kit
Enables the fixation and permeabilization of cells which is necessary for staining intracellular cytokines with fluorochrome-conjugated anti-cytokine antibodies
BD Fetal Bovine Serum Stain Buffer
For the immunofluorescent staining of single-cell suspensions
BD Phosflow Lyse/Fix Buffer 5X
For the lysing and fixing of whole blood for use with intracellular flow cytometry
BD Bovine serum albumin Stain Buffer
For immunofluorescent staining of single-cell suspensions
BD Phosflow Perm Buffer II and Perm Buffer III
For the permeabilization of fixed Peripheral Blood Mononuclear Cells (PBMCs) or lysed whole blood
BD Cytometric Bead Array Human Th1/Th2/Th17 Kit
Optimized for analysis of physiologically relevant concentrations (pg/mL levels) of specific cytokine proteins in tissue culture supernatants, EDTA plasma, and serum samples
BD APC BrdU Flow Kit
Comes with 65μL of APC-conjugated anti-BrdU antibody
BD Lysing Buffer
Buffered ammonium chloride-based lysing reagent
BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution Kit with BD GolgiStop™
Provides GolgiStop protein transport inhibitor containing monensin
eBioscience™ Flow Cytometry Staining Buffer Non-distribution product as customer accommodation.
Designed for use in immunofluorescent staining protocols of cells in suspension. Flow Cytometry Staining Buffer is a buffered saline solution containing fetal bovine serum and sodium azide (0.09%) as a preservative.
Coulter™ Cyto-Trol™ Control Cells
Assess the activity of monoclonal antibodies by flow cytometry
Molecular Probes™ Click-iT™ Plus EdU Alexa Fluor™ 647 Flow Cytometry Assay Kit Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
Click-iT™ Plus EdU Alexa Fluor™ 647 Flow Cytometry Assay Kit
BD Cytometric Bead Array (CBA) Mouse/Rat Soluble Protein Master Buffer Kit
Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
BD Human Soluble Protein Master Buffer Kit
Along with Human soluble protein flex set system buffer kit employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
Coulter™ Flow-Check™ Fluorospheres
Used to verify instrument optical alignment and fluidic functionality
Coulter™ IsoFlow™ Sheath Fluid
Nonfluorescent, balanced electrolye solution for use on Coulter flow cytometers with light scatter and fluorescent applications
R&D Systems™ Flow Cytometry Staining Buffer (1X) Available on GSA/VA Contract for Federal Government customers only.
Designed for flow cytometry applications with nonconjugated or fluorochrome-conjugated antibodies
BD Sphero™ Fluorescent Particles
For flow cytometry calibration and alignment
R&D Systems™ Flow Cytometry Fixation Buffer (1X) Available on GSA/VA Contract for Federal Government customers only.
Optimized for fixation and storage of single-cell suspensions intended for subsequent intracellular flow cytometry applications
BD MitoScreen Flow Cytometry Mitochondrial Membrane Potential Detection Kit
Contains JC-1, which has been optimized for use in flow cytometry, and 10 assay buffer
R&D Systems™ Flow Cytometry Mouse Lyse Buffer (10X)
Optimized to prepare mouse samples containing an excess of red blood cells, including spleen or whole blood samples
BD PI/RNase Staining Buffer
May be used to analyze cell cycle by flow cytometry or used with antibodies to examine the expression of proteins during the cell cycle
BD Cytometric Bead Array (CBA) Functional Bead Conjugation Buffer Set
Used to covalently link watersoluble proteins to surface of color-coded functional beads
R&D Systems™ Flow Cytometry Permeabilization/Wash Buffer I (1X) Available on GSA/VA Contract for Federal Government customers only.
Optimized for permeabilization of single-cell suspensions, dilution and washing of these samples for subsequent intracellular flow cytometry applications
Coulter™ CYTO-COMP™ Quality Control Kits
Used to establish and monitor color compensation settings for multicolor analysis
Coulter™ Whole Blood Lysing Reagent Kit
For the preparation of whole blood, bone marrow aspirates, and other sample types for analysis
Coulter™ Immunochemistry Reagents
Designed to support fluorescence microscopy and flow cytometry
  spinner