Fluorometers
Invitrogen™ Qubit™ Assay Tubes Non-distribution product as customer accommodation.
For use with the Qubit 2.0 Fluorometer
Invitrogen™ Qubit™ 4 Fluorometer Non-distribution product as customer accommodation.
The Qubit 4 Flurometer is the latest version of the popular Qubit fluorometer designed to accurately measure DNA, RNA, and protein quantity, and now also RNA integrity and quality, using the highly sensitive Qubit assays.
TECHREF™ Type RP-1118 Thermal Printer Paper
Used with HORIBA SLFA single load and turntable XRF analyzers.
Invitrogen™ Qubit™ 4 NGS Starter Kit Non-distribution product as customer accommodation.
For use with the Qubit 4 Flurometer.
Invitrogen™ Qubit™ 4 Quantitation Starter Kit Non-distribution product as customer accommodation.
For use with the Qubit 4 Flurometer.
Invitrogen™ Qubit™ 1X dsDNA HS Assay Kit Non-distribution product as customer accommodation.
The Qubit 1X dsDNA HS (High-Sensitivity) Assay Kit is designed specifically for use with the Qubit Fluorometer.
Invitrogen™ Qubit™ 4 RNA IQ Starter Kit Non-distribution product as customer accommodation.
For use with the Qubit 4 Flurometer.
Promega Quantus™ Fluorometer
Dual-channel fluorometer for personal quantitation workflow. Designed to provide highly sensitive fluorescent detection when quantifying nucleic acids, the compact instrument is simple to operate. The Quantus Fluorometer is optimized with preprogrammed settings for Promega QuantiFluor™ Dye Systems to quantitate nucleic acids and offers the flexibility to create customized methods and quantitation settings for other fluorescent dyes.
Invitrogen™ Qubit™ RNA IQ Assay Kit Non-distribution product as customer accommodation.
The Qubit RNA IQ Assay Kit provides a fast, simple method to check whether an RNA sample has degraded using the Qubit 4 Fluorometer (required). The assay utilizes two unique dyes: one that binds to large, intact and/or highly structured RNA (mRNA, tR
Thermo Scientific™ NanoDrop™ 3300 Fluorospectrometer Available on GSA/VA Contract for Federal Government customers only.
Choose this versatile, cost-effective fluorospectrometer for broad spectrum fluorescence analysis.
Promega QuantiFluor Service Agreement, 1 each
Service Agreement for QuantiFluor-ST and QuantiFluor-P Fluorometer Instruments. The instruments are discontinued. Service available until March 1, 2019.
Promega™ Solid Standard for QuantiFluor™-ST Fluorometer
Designed to verify the stability of the fluorometer by checking drift of the calibration.
Promega™ QuantiFluor™-ST and QuantiFluor-P Single-Tube Fluorometers: PCR Tube Adapter
PCR Tube Adapter for QuantiFluor single-tube fluorometers
Promega Quantus Instrument Standard Service Agreement, 1 each
The Quantus Fluorometer 1-year service agreement.
Promega™ Replacement AC Adapter for QuantiFluor™-ST
For use with the QuantiFluor-ST Single-Tube Fluorometer.
TECHREF™ Type AT-700 Assembly Tool
Designed and built to ease the assebmley of the TECHREF Type C-700 sample cells.
  spinner