Fluorometers and Fluorescence Polarization Systems
Invitrogen™ Qubit™ 4 Fluorometer Non-distribution product as customer accommodation.
The Qubit 4 Flurometer is the latest version of the popular Qubit fluorometer designed to accurately measure DNA, RNA, and protein quantity, and now also RNA integrity and quality, using the highly sensitive Qubit assays.
Invitrogen™ Qubit™ 4 Quantitation Starter Kit Non-distribution product as customer accommodation.
For use with the Qubit 4 Flurometer.
Invitrogen™ Qubit™ 1X dsDNA HS Assay Kit Non-distribution product as customer accommodation.
The Qubit 1X dsDNA HS (High-Sensitivity) Assay Kit is designed specifically for use with the Qubit Fluorometer.
Invitrogen™ Qubit™ 4 NGS Starter Kit Non-distribution product as customer accommodation.
For use with the Qubit 4 Flurometer.
Promega Quantus™ Fluorometer
Dual-channel fluorometer for personal quantitation workflow. Designed to provide highly sensitive fluorescent detection when quantifying nucleic acids, the compact instrument is simple to operate. The Quantus Fluorometer is optimized with preprogrammed settings for Promega QuantiFluor™ Dye Systems to quantitate nucleic acids and offers the flexibility to create customized methods and quantitation settings for other fluorescent dyes.
Invitrogen™ Qubit™ RNA IQ Assay Kit Non-distribution product as customer accommodation.
The Qubit RNA IQ Assay Kit provides a fast, simple method to check whether an RNA sample has degraded using the Qubit 4 Fluorometer (required). The assay utilizes two unique dyes: one that binds to large, intact and/or highly structured RNA (mRNA, tR
Thermo Scientific™ NanoDrop™ 3300 Fluorospectrometer Available on GSA/VA Contract for Federal Government customers only.
Choose this versatile, cost-effective fluorospectrometer for broad spectrum fluorescence analysis.
Promega™ QuantiFluor™-ST and QuantiFluor-P Single-Tube Fluorometers: PCR Tube Adapter
PCR Tube Adapter for QuantiFluor single-tube fluorometers
Invitrogen™ Qubit™ 4 RNA IQ Starter Kit Non-distribution product as customer accommodation.
For use with the Qubit 4 Flurometer.
  spinner