Microinjectors for Molecular Biology
Eppendorf™ Femtotips™ Microloader Tips for Femtojet Microinjector Available on GSA/VA Contract for Federal Government customers only.
Tips to fill microinjection capillaries and recovering surplus solution from capillary
Eppendorf™ Femtotips™ Microinjection Capillary Tips Available on GSA/VA Contract for Federal Government customers only.
Disposable glass capillaries for reproducible microinjection into adherent and suspension cells
Eppendorf™ FemtoJet™ 4i Microinjector Available on GSA/VA Contract for Federal Government customers only.
The electronic microinjector, FemtoJet 4i, with its simple operation and wide range of functionality is ideally suited for injecting small to intermediate volumes of up to 100 pL into adherent cells and suspension cells for research purposes.
Eppendorf™ FemtoJet™ 4x Microinjector Available on GSA/VA Contract for Federal Government customers only.
The FemtoJet 4x is the ideal solution if your research demands injection volumes greater than 100 pL and/or longer series at higher pressures. Ideal for use in functional genomics and developmental biology applications.
  spinner