Plates Prepared Microbiology Media
Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium Available on GSA/VA Contract for Federal Government customers only.
Grow a wide variety of microorganisms on this general purpose non-selective medium that includes sheep blood to promote haemolysis.
Thermo Scientific™ MacConkey Agar Medium Available on GSA/VA Contract for Federal Government customers only.
Detect and enumerate coliform organisms, E. coli, while isolating and detecting Salmonella and Shigella spp. in both pathological and food specimens.
Thermo Scientific™ Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate Available on GSA/VA Contract for Federal Government customers only.
Grow fastidious organisms, detect and enumerate coliforms, and isolate and detect Salmonella and Shigella spp. on a single plate.
Thermo Scientific™ Columbia CNA Agar with Sheep Blood Medium Available on GSA/VA Contract for Federal Government customers only.
Isolate Gram-positive organisms from clinical samples with this selective media traditionally used to identify staphylococci, streptococci, and S. pneumoniae.
Thermo Scientific™ Remel™ Middlebrook 7H11 Agar Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Middlebrook 7H11 Agar is used for the cultivation and isolation of Mycobacterium species.
Thermo Scientific™ Chocolate Agar Medium Available on GSA/VA Contract for Federal Government customers only.
Obtain fast and accurate results with a nutritious medium for the isolation of fastidious organisms Neisseria, Haemophilus, and Gardnerella vaginalis.
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (TSA II™) with Sheep Blood Available on GSA/VA Contract for Federal Government customers only.
Used for cultivating fastidious microorganisms and for visualization of hemolytic reactions produced by many bacterial species
Thermo Scientific™ Spectra™ MRSA Medium Available on GSA/VA Contract for Federal Government customers only.
Obtain fast and accurate results with a selective chromogenic medium for screening clinical samples for the presence of Methicillin Resistant Staphylococcus aureus.
Thermo Scientific™ Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
BD BBL™ Prepared Plated Media: MacConkey II Agar Available on GSA/VA Contract for Federal Government customers only.
Selective and differential media for the detection of coliform organisms and enteric pathogens
BD BBL™ Prepared Plated Media: CHROMagar™ MSRA II
For the qualitative direct detection of nasal colonization by methicillin-resistant Staphylococcus aureus (MRSA)
Thermo Scientific™ Spectra™ VRE Medium Available on GSA/VA Contract for Federal Government customers only.
Obtain fast and accurate results with a chromogenic medium designed for screening clinical samples for the presence of vancomycin-resistant Enterococcus (VRE).
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (Soybean-Casein Digest Agar) Available on GSA/VA Contract for Federal Government customers only.
Used for isolation and cultivation of non-fastidious and fastidious microorganisms
Thermo Scientific™ Remel™ D/E (Dey-Engley) Neutralizing Broth Available on GSA/VA Contract for Federal Government customers only.
Use in environmental sampling procedures requiring neutralization of chemical disinfectants to distinguish bacteriostatic and bactericidal activity.
Thermo Scientific™ Remel™ HE Agar (Hektoen Enteric Agar) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel HE Agar (Hektoen Enteric Agar) facilitates isolation and differentiation of gram-negative enteric microorganisms.
Thermo Scientific™ Brilliance™ CampyCount Agar Available on GSA/VA Contract for Federal Government customers only.
Accurately and efficiently identify and enumerate Campylobacter jejuni and C. coli from poultry samples on plate reader friendly, chromogenic plates.
BD BBL™ Prepared Plated Media: Chocolate II Agar (GC II Agar with Hemoglobin and IsoVitaleX™) Available on GSA/VA Contract for Federal Government customers only.
For the isolation and cultivation of fastidious microorganisms, especially Neisseria and Haemophilus species, from a variety of clinical specimens
BD BBL™ Prepared Plated Media: Columbia CNA Agar with 5% Sheep Blood Available on GSA/VA Contract for Federal Government customers only.
Selective and differential media used for the isolation and differentiation of gram-positive microorganisms from clinical and nonclinical materials
Thermo Scientific™ Remel™ Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Microbial Content Test Agar) Available on GSA/VA Contract for Federal Government customers only.
Use for detection and enumeration of microorganisms present on surfaces of sanitary importance.
BD BBL™ Trypticase™ Soy Agar w/5% Sheep Blood/ MacConkey II, I Plate™ Prepared Media Available on GSA/VA Contract for Federal Government customers only.
Two media formulations in single plate for cultivating fastidious microorganisms
Thermo Scientific™ Columbia Blood Agar with Sheep Blood Medium Available on GSA/VA Contract for Federal Government customers only.
Isolate and cultivate fastidious microorganisms with clearly visible haemolytic reactions (staphylococci and streptococci) with a medium containing sheep blood.
BD BBL™ Prepared Sterile Pack Settling Plated Trypticase™ Soy Agar
Plated media in gamma-irradiated packs for isolator systems-multiwrap packaging protects media from vaporized hydrogen peroxide exposure
BD BBL™ Prepared Plated Media: Mueller Hinton II Agar Available on GSA/VA Contract for Federal Government customers only.
For antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method
Thermo Scientific™ Remel™ Cetrimide Selective Agar Available on GSA/VA Contract for Federal Government customers only.
Use for selective isolation and presumptive identification of Pseudomonas aeruginosa and other gram-negative, non-fermentative bacteria.
Thermo Scientific™ Remel Mitchison 7H11 Selective Agar w/Carbenicillin, Polymyxin B, Amphotericin B, Trimethoprim Available on GSA/VA Contract for Federal Government customers only.
The Thermo Scientific™ Remel Mitchison 7H11 Selective Agar with antibiotics provides selective isolation of mycobacteria from specimens containing mixed flora.
BD BBL™ Trypticase™ Soy Agar with Lecithin and Polysorbate 80 Sterile Pack Media - RODAC Plates
Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure
BD Difco™ Buffered Listeria Enrichment Broth Base
Enrichment broth for cultivation of Listeria spp. from food
Thermo Scientific™ Remel™ Burkholderia Cepacia Selective Agar Available on GSA/VA Contract for Federal Government customers only.
For selective and differential isolation of Burkholderia cepacia complex from respiratory secretions of patients with cystic fibrosis.
Thermo Scientific™ Remel™ Mannitol Salt Agar (MSA) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Mannitol Salt Agar (MSA) is for the selective isolation, differentiation, and enumeration of staphylococci.
  spinner