Human Plasmas
EMD Millipore™ Calbiochem™ Human γ-Globulin, Serum Available on GSA/VA Contract for Federal Government customers only.
Temporarily boosts patient's immunity against disease
EMD Millipore™ Calbiochem™ Albumin, Human Serum, 30% Aqueous Solution Available on GSA/VA Contract for Federal Government customers only.
Suitable for use as Protein base or filler Antigen
EMD Millipore™ Calbiochem™ Protein C, Activated, Human Plasma
Native, activated protein C from human plasma
EMD Millipore™ Calbiochem™ Protein C, Human Plasma
Potent anticoagulant that acts through selective inactivation of Factors Va and VIIIa
EMD Millipore™ Calbiochem™ Human IgG, Fc Fragment, Plasma Available on GSA/VA Contract for Federal Government customers only.
Native IgG Fc fragment derived from normal human plasma IgG
EMD Millipore™ Calbiochem™ Albumin, Human Serum, Fraction V, Low Heavy Metals Available on GSA/VA Contract for Federal Government customers only.
Prepared under non-denaturing conditions by modification of Cohn procedure
EMD Millipore™ Calbiochem™ Human IgA, Plasma Available on GSA/VA Contract for Federal Government customers only.
Native IgA purified from human plasma
EMD Millipore™ Calbiochem™ Complement C1q-Depleted Serum, Human Available on GSA/VA Contract for Federal Government customers only.
For detection and quantitation of hemolytically active complement components
EMD Millipore™ Calbiochem™ Complement C3-Depleted Serum, Human
For detection and quantitation of hemolytically active complement components
EMD Millipore™ Calbiochem™ Complement C5-Depleted Serum, Human Available on GSA/VA Contract for Federal Government customers only.
For detection and quantitation of hemolytically active complement components
EMD Millipore™ Calbiochem™ Complement C9-Depleted Serum, Human
For detection and quantitation of hemolytically active complement components
EMD Millipore™ Calbiochem™ Human IgG, Fab Fragment, Plasma
Native IgG Fab fragment derived from normal human plasma IgG
EMD Millipore™ Calbiochem™ Human IgD, Myeloma Plasma
Native IgD from human myeloma plasma
EMD Millipore™ Calbiochem™ Prealbumin, Human Plasma Available on GSA/VA Contract for Federal Government customers only.
Transports retinol-binding protein, thyroxine, and vitamin A
EMD Millipore™ Calbiochem™ Protein S, Human Plasma
Functions as cofactor for anticoagulant activity of activated protein C
EMD Millipore™ Calbiochem™ Platelet Membrane Glycoproteins, IIbIIIa, Human Platelets Available on GSA/VA Contract for Federal Government customers only.
Native, platelet membrane glycoprotein IIbIIIaA from human platelets
EMD Millipore™ Calbiochem™ Albumin, Human Serum, Non-denatured Available on GSA/VA Contract for Federal Government customers only.
Suitable for use as Antigen, Culture media for animal cells, Protein standard
EMD Millipore™ Calbiochem™ Albumin, Human Serum, Fraction V Available on GSA/VA Contract for Federal Government customers only.
Prepared under non-denaturing conditions by modification of Cohn procedure
  spinner